Skip to Content
Merck
All Photos(3)

Key Documents

HPA042199

Sigma-Aldrich

Anti-GPSM1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AGS3, Anti-DKFZP727I051, Anti-G-Protein Signaling Modulator 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SPAASEKPDLAGYEAQGARPKRTQRLSAETWDLLRLPLEREQNGDSHHSGDWRGPSRDSLPLPVRSRK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPSM1(26086)

General description

G-protein-signaling modulator 1 (GPSM1) is also called activator of G protein signalling 3 (AGS3). The GPSM1 gene is mapped to human chromosome 9q34.3. AGS3 has a N-terminal domain with seven tetratri-copeptides (TPR) repeats and the C-terminal domain with four G-protein regulatory motifs (GPR) or GoLocomotifs. Brain and testes express AGS3 in high levels.

Immunogen

G-protein signaling modulator 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

G-protein-signaling modulator 1 (GPSM1) or activator of G protein signalling 3 (AGS3) functions to regulate G protein signalling. AGS3 functions to orient mitotic spindle and modulates development of cerebral cortical progenitor cells during neurogenesis. The GPR motif of AGS3 cytokine may regulate cytokine production in airway inflammation. It also regulates autophagic pathway and protein trafficking. AGS3 is down regulated human esophageal squamous cell carcinoma. AGS3 is also less expressed in multiple myeloma. Abnormal expression of AGS3 in renal epithelium is implicated in polycystic kidney disease (PKD).(10) Defects in AGS3 gene locus is implicated in MORM syndrome (mental retardation, truncal obesity, retinal dystrophy and micropenis).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST82026

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A role for activator of G-protein signaling 3 (AGS3) in multiple myeloma
Shao S, et al.
International Journal of Hematology, 99(1), 57-68 (2014)
MORM syndrome (mental retardation, truncal obesity, retinal dystrophy and micropenis), a new autosomal recessive disorder, links to 9q34
Hampshire D, et al.
European Journal of Human Genetics, 14(5), 543-543 (2006)
G protein betagamma subunits and AGS3 control spindle orientation and asymmetric cell fate of cerebral cortical progenitors
Sanada K and Tsai L
Cell, 122(1), 119-131 (2005)
AGS proteins, GPR motifs and the signals processed by heterotrimeric G proteins
Lanier SM
Biology of the Cell, 96(5), 369-372 (2004)
The G-protein regulator AGS3 controls an early event during macroautophagy in human intestinal HT-29 cells
Pattingre S, et al.
The Journal of Biological Chemistry (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service