Skip to Content
Merck
All Photos(7)

Documents

HPA038225

Sigma-Aldrich

Anti-VGLL4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-KIAA0121, Anti-Vestigial like 4 (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PISPSKRKFSMEPGDEDLDCDNDHVSKMSRIFNPHLNKTANGDCRRDPRERSRSPIERAVAPTMSLHGSHLYTSLP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VGLL4(9686)

Related Categories

General description

Vestigial-like 4 (VGLL4) belongs to the family of VGLL proteins and is characterized by two TDU domains at the C-terminal, unlike other members of the family. Vgl-4 is highly expressed in the nucleus.

Immunogen

vestigial-like family member 4

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry. The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Vestigial-like 4 (VGLL4) stimulates apoptotic pathway by negatively regulating inhibitor of apoptosis proteins (IAPs). The encoded protein acts as a potential tumor suppressor and prognostic marker in gastric cancer and lung cancer. Knockdown of VGLL4 expression leads to progression of esophageal squamous cell carcinoma (ESCC).Therefore, restoring the function of VGLL4 can be a potential method for treating ESCC. Experimental studies states that VGLL4 is involved in regulation of cell mobility, migration and invasion.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79374

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

VGLL4 inhibits EMT in part through suppressing Wnt/?-catenin signaling pathway in gastric cancer.
Li H
Medical Oncology (Northwood, London, England), 32 (2015)
A novel inhibitor of apoptosis protein (IAP)-interacting protein, Vestigial-like (Vgl)-4, counteracts apoptosis-inhibitory function of IAPs by nuclear sequestration.
Jin HS
Biochemical and Biophysical Research Communications, 412, 454-459 (2011)
VGLL4 functions as a new tumor suppressor in lung cancer by negatively regulating the YAP-TEAD transcriptional complex.
Zhang W
Cell Research, 24, 331-331 (2014)
A peptide mimicking VGLL4 function acts as a YAP antagonist therapy against gastric cancer.
Jiao S
Cancer Cell, 25, 166-180 (2014)
Downregulation of VGLL4 in the progression of esophageal squamous cell carcinoma.
Jiang W
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 36, 1289-1297 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service