Skip to Content
Merck
All Photos(4)

Key Documents

HPA032045

Sigma-Aldrich

Anti-CLNS1A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CLCI, Anti-ICln

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SFLKSFPPPGPAEGLLRQQPDTEAVLNGKGLGTGTLYIAESRLSWLDGSGLGFSLEYPTISLHALSRDRSDCLGEHLYVMVNAKFEEESKEPVADEE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLNS1A(1207)

General description

Chloride nucleotide-sensitive channel 1A (CLNS1A), also called ICLN, is a 25 kDa channel protein with nucleotide and calcium binding regions. The CLNS1A gene is located on human chromosome 11q14.1.

Immunogen

chloride channel, nucleotide-sensitive, 1A

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Chloride nucleotide-sensitive channel 1A (CLNS1A) interacts with serine/threonine-protein kinase, RioK1 and mediates recruitment of spliceosomal Sm proteins for methylation. CLNS1A controls the quality ofsmall nuclear ribonucleoprotein (Sm) oligomers. It is part of 6S complex and 20S methylosome complex. CLNS1A acts as a chaperone for Sm-specific assembly and regulates snRNP biogenesis. Mutations inCLNS1A may impair nucleotide binding and kinetics.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78700

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toward an Assembly Line for U7 snRNPs INTERACTIONS OF U7-SPECIFIC Lsm PROTEINS WITH PRMT5 AND SMN COMPLEXES
Azzouz TN, et al.
The Journal of Biological Chemistry, 280(41), 34435-34440 (2005)
New mammalian chloride channel identified by expression cloning
Paulmichl M, et al.
Nature, 356(6366), 238-238 (1992)
RioK1, a new interactor of protein arginine methyltransferase 5 (PRMT5), competes with pICln for binding and modulates PRMT5 complex composition and substrate specificity
Guderian G, et al.
The Journal of Biological Chemistry, 286(3), 1976-1986 (2011)
The methylosome, a 20S complex containing JBP1 and pICln, produces dimethylarginine-modified Sm proteins
Friesen WJ, et al.
Molecular and Cellular Biology, 21(24), 8289-8300 (2001)
pICln inhibits snRNP biogenesis by binding core spliceosomal proteins
Pu WT, et al.
Molecular and Cellular Biology, 19(6), 4113-4120 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service