Skip to Content
Merck
All Photos(6)

Key Documents

HPA027366

Sigma-Aldrich

Anti-HTRA2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-High temperature requirement protein A2, Anti-HtrA2, Anti-Omi stress-regulated endoprotease, Anti-Serine protease 25, Anti-Serine protease HTRA2, mitochondrial, Anti-Serine proteinase OMI

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

MAAPRAGRGAGWSLRAWRALGGIRWGRRPRLTPDLRALLTSGTSDPRARVTYGTPSLWARLSVGVTEPRACLTSGTPGPRAQLTAVTPDTRTREASENSGTRSR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HTRA2(27429)

General description

The gene HTRA2 (high temperature requirement protein A2) is mapped to human chromosome 2p13. The encoded protein has a PDZ (postsynaptic density of 95 kDa, disc large and zonula occludens 1) domain and a protease domain. It is mainly present in the mitochondrial intermembrane space and goes to the cytoplasm in presence of cellular stress.

Immunogen

Serine protease HTRA2, mitochondrial Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-HTRA2 antibody produced in rabbit has been used in western blotting.[1]

Biochem/physiol Actions

HTRA2 (high temperature requirement protein A2) is a mitochondrial serine protease and is associated with various neurodegenerative disorders. HTRA proteases recognize the exposed hydrophobic areas in proteins and are responsible for their degradation. It helps in mitochondrial homeostasis and plays a proapoptotic role in the cytoplasm. It is also involved in the development of cancer. It might be associated with various cancers due to its apoptotic function.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70804

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lyme disease spirochaetes possess an aggrecan-binding protease with aggrecanase activity.
Russell TM and Johnson BJ
Molecular Microbiology, 90, 228-240 (2013)
Intra- and intersubunit changes accompanying thermal activation of the HtrA2(Omi) protease homotrimer.
Jarzab M, et al.
Biochimica et Biophysica Acta, 1864, 283-296 (2016)
High-temperature-required protein A2 as a predictive marker for response to chemotherapy and prognosis in patients with high-grade serous ovarian cancers.
Miyamoto M, et al.
British Journal of Cancer, 112, 739-744 (2015)
Parkinson's disease: a genetic perspective.
Belin AC and Westerlund M
FEBS Journal, 275, 1377-1383 (2008)
Stress Conditions Increase Vimentin Cleavage by Omi/HtrA2 Protease in Human Primary Neurons and Differentiated Neuroblastoma Cells.
Lucotte B, et al.
Molecular Neurobiology, 52, 1077-1092 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service