Skip to Content
Merck
All Photos(4)

Key Documents

HPA026863

Sigma-Aldrich

Anti-CYP1B1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CP1B, Anti-GLC3A, Anti-cytochrome P450, family 1, subfamily B, polypeptide 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

SSFVPVTIPHATTANTSVLGYHIPKDTVVFVNQWSVNHDPLKWPNPENFDPARFLDKDGLINKDLTSRVMIFSVGKRRCIGEELSKMQLFLFISILAHQCDFRANPNEPAKMNFSYGLTIKPKSFKVNVTLRESMELLDSAVQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYP1B1(1545)

General description

The Cytochrome P4501B1 (CYP1B1) gene with three exons and two introns is mapped to human chromosome 2p21-22. The 543 amino acid protein encoded by this gene is expressed mainly in the adrenals, ovary. The protein is a member of cytochrome P4501, a subfamily of CYP1 family .
CYP1B1 is deprived of a consensus TATA box in the promoter region and it is characterized with nine tetrachlorodibenzo-p-dioxin (TCDD) responsive enhancer core binding motifs present within a 2.5kb pair genomic fragment of the transcription initiation start site.

Immunogen

cytochrome P450, family 1, subfamily B, polypeptide 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Tissue distribution pattern analysis shows that cytochrome P4501B1 (CYP1B1) plays a vital role in metabolism of xenobiotics and possibly steroid hormones. CYP1B1 might be responsible for the metabolism of biologically active molecule associated with eye development. Any truncating mutation of the gene coding for this protein causes primary congenital glaucoma. Isoflavone genistein stimulates the gene expression of CYP1B1, which plays a vital role in human breast cancer development and progression. CYP1B1 enzyme catalyzes the conversion of pro-carcinogens to carcinogens and also facilitates estrogen metabolism. Single nucleotide polymorphism (SNP) of CYP1B1 might enhance susceptibility and pathological development of hepatocellular carcinoma (HCC) in Chinese population.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86682

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genistein Induces Cytochrome P450 1B1 Gene Expression and Cell Proliferation in Human Breast Cancer MCF-7 Cells.
Wei YK
Journal of Environmental Pathology, Toxicology and Oncology, 34(2), 153-159 (2015)
Identification of three different truncating mutations in cytochrome P4501B1 (CYP1B1) as the principal cause of primary congenital glaucoma (Buphthalmos) in families linked to the GLC3A locus on chromosome 2p21.
Human Molecular Genetics, 6(4), 641-647 (1997)
Polymorphisms of the CYP1B1 gene and hepatocellular carcinoma risk in a Chinese population.
Liu F
Gene, 564(1), 14-20 (2015)
Isolation and characterization of the human cytochrome P450 CYP1B1 gene.
Tang YM
The Journal of Biological Chemistry, 271(45), 28324-28330 (1996)
Mateusz Kurzawski et al.
Pharmacological reports : PR, 64(4), 927-939 (2012-10-23)
Expression of drug-metabolizing enzymes and drug transporters in liver is mainly regulated by a system of nuclear receptors. The aim of the current study was to investigate the expression of nuclear receptors, as well as these enzymes and transporters, in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service