Skip to Content
Merck
All Photos(3)

Key Documents

HPA026763

Sigma-Aldrich

Anti-NENF antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cell immortalization-related protein 2, Anti-Neudesin, Anti-Neuron-derived neurotrophic factor, Anti-SPUF protein, Anti-Secreted protein of unknown function

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

RLFTEEELARYGGEEEDQPIYLAVKGVVFDVTSGKEFYGRGAPYNALTGKDSTRGVAKMS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NENF(29937)

Related Categories

General description

Neuron derived neurotrophic factor (NENF), also called as neudesin and GIG47, is mapped to human chromosome 1p33. This gene is expressed only in vertebrates. NENF codes for a novel secretory protein with 172 amino acids named neudesin. Primary structure of this protein might contain cytochrome b5-like heme/steroid-binding domain. In mouse, neudesin is expressed in heart, kidney, brain, and lung.

Immunogen

Neudesin Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Neuron derived neurotrophic factor (NENF) or GIG47 as an oncogene plays a crucial role in human tumorigenesis. It is highly expressed in various human cancer tissues, such as uterine cervix cancer, lymphoma, colon, lung, skin, leukemia and even breast cancer. Neudesin acts as a neurotrophic factor in neurons and undifferentiated neural cells. The neurotrophic property of this protein is transmitted by the activation of mitogen-activated protein (MAP) and phosphatidylinositol 3-kinase (PI-3K) pathways. NENF or neudesin is a primary extracellular heme-binding protein implicated in intercellular signal transduction. In mouse, neudesin plays a vital role in neural cell proliferation and neuronal differentiation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76694

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The functional and structural characterization of a novel oncogene GIG47 involved in the breast tumorigenesis.
The functional and structural characterization of a novel oncogene GIG47 involved in the breast tumorigenesis.
Han KH
BMC Cancer, 12:274 (2012)
Neurotrophic activity of neudesin, a novel extracellular heme-binding protein, is dependent on the binding of heme to its cytochrome b5-like heme/steroid-binding domain.
Kimura I
The Journal of Biological Chemistry, 283(7), 4323-4331 (2008)
Neudesin, a secreted factor, promotes neural cell proliferation and neuronal differentiation in mouse neural precursor cells.
Kimura I
Journal of Neuroscience Research, 83(8), 1415-1424 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service