Skip to Content
Merck
All Photos(8)

Key Documents

HPA026744

Sigma-Aldrich

Anti-SPACA1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Sperm acrosomal membrane-associated protein 32, Anti-Sperm acrosome membrane-associated protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200-1:500
western blot: 0.04-0.4 μg/mL

immunogen sequence

REVILTNGCPGGESKCVVRVEECRGPTDCGWGKPISESLESVRLACIHTSPLNRFKYMWKL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SPACA1(81833)

General description

Sperm acrosome associated 1(spaca1) belongs to the family of globozoospermia-related genes. This protein is present in the equatorial segment of mammalian spermatozoa. The gene coding for spaca1 protein is mapped to human chromosome 6q15. Its expression is found to be in sync with acrosome development in human testicular cells.

Immunogen

Sperm acrosome membrane-associated protein 1 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Sperm acrosome associated 1(spaca1) plays a vital role in sperm-egg fusion. Proteomic analysis states that spaca1 concentration is decreased in globozoospermia patients. The encoded protein can be regulated by phosphorylation of conserved sequence atY268 tyrosine phosphorylation site.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77394

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guy Longepied et al.
Human reproduction (Oxford, England), 25(10), 2655-2663 (2010-08-19)
Deletion of the entire AZFb interval from the Y chromosome is strictly associated with azoospermia arising from maturation arrest during meiosis. Here, we describe the exceptional case of an oligozoospermic man, 13-1217, with an AZFb + c (P5/distal-P1) deletion. Through
Eric Streichemberger et al.
Human reproduction (Oxford, England), 27(7), 1939-1945 (2012-04-18)
AZFc deletions of the Y chromosome are the major known genetic cause of spermatogenetic failure. Meiotic studies have shown a prevalence of synaptonemal complex fragmentation and an excess of early-stage sperm cells, suggesting that the maturation block could involve apoptosis.
Marine Paci et al.
Reproductive biomedicine online, 35(5), 562-570 (2017-09-09)
The aim of this study was to characterize the nuclear lamina (NL) and lamin chromatin-partners in spermatozoa from four DPY19L2-deleted globozoospermic patients. We tested for spermatid transcripts encoding lamins and their chromatin-partners emerin, LAP2α, BAF and BAF-L, by reverse transcriptase-PCR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service