Skip to Content
Merck
All Photos(2)

Key Documents

HPA026110

Sigma-Aldrich

Anti-MUC15 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MUC-15, Anti-Mucin-15

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

KENQDINTTQNIAEVFKTMENKPISLESEANLNSDKENITTSNLKASHSPPLNLPNNSHGITDFSSNSSAEHSLGSLKPTSTISTSPPLIHSFVSKVPWNAPIADEDLLPISAHPNATPALSSENFTWSLVNDTVKTPDNSSITVSILSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUC15(143662)

General description

Mucin-15 (MUC15) is a type-I membrane-bound glycoprotein. The gene encoding it is localized on human chromosome 11p14.3.

Immunogen

Mucin-15 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Mucin-15 (MUC15) is associated with colorectal and gastric cancers. It has been shown to interact with epidermal growth factor receptor (EGFR) and may aid in its degradation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70006

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shunda Wang et al.
Medical science monitor : international medical journal of experimental and clinical research, 26, e926432-e926432 (2020-10-15)
BACKGROUND MUC15, one of the hydrophilic glycoproteins that protect wet-surfaced epithelia, has been shown to be involved in tumorigenesis of various tumors. However, the mechanism of MUC15 in pancreatic cancer have not been revealed yet. Our study focused on investigating
John Huang et al.
Carcinogenesis, 30(8), 1452-1458 (2009-06-13)
Mucins play a key role in tumorigenesis. MUC15 is a membrane-bound mucin and the MUC15 messenger RNA (mRNA) has been detected in various organs. However, its role in tumor malignancy is still unclear. This study was to investigate the MUC15
Hye Rim Oh et al.
Pathology oncology research : POR, 21(3), 713-718 (2015-01-13)
Mucins are important in tumorigenesis and expressional alterations of mucins are common in human cancers. A membrane-bound mucin MUC15 and secreted mucins MUC4 and MUC7 are known to involve in tumorigenesis, but their mutation status in cancers remains unknown. Aim
Ruo-Yu Wang et al.
Gastroenterology, 145(6), 1436-1448 (2013-08-13)
Aberrant expression of MUC15 correlates with development of colorectal adenocarcinoma, and MUC15 has been reported to prevent trophoblast invasion of human placenta. However, little is known about the role of MUC15 in pathogenesis of hepatocellular carcinoma (HCC). We analyzed HCC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service