Skip to Content
Merck
All Photos(5)

Documents

HPA026089

Sigma-Aldrich

Anti-SLC27A2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FATP-2, Anti-Solute carrier family 27 member 2, Anti-THCA-CoA ligase, Anti-VLACS, Anti-VLCS, Anti-Very long-chain acyl-CoA synthetase, Anti-Very long-chain-fatty-acid-CoA ligase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

SLLHCFQCCGAKVLLVSPELQAAVEEILPSLKKDDVSIYYVSRTSNTDGIDSFLDKVDEVSTEPIPESWRSEVTFSTPALYIYTSGTTGLPKAAMITHQRIWYGTGLTFVSGLKADDVIYITLPFYHSAALLIGIHGCIVAGAT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC27A2(11001)

General description

Solute carrier family 27 member 2 (SLC27A2) is expressed in microsomes and peroxisomes. The gene encoding it is localized on human chromosome 15q21.2. It belongs to the fatty acid transport protein family.

Immunogen

Very long-chain acyl-CoA synthetase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Solute carrier family 27 member 2 (SLC27A2) has a role in fatty acid transport. It also mediates the activation of fatty acids having a chain length of 22 or more carbons. It is mainly responsible for the production of CoA (coenzyme A) derivatives of n-3 fatty acids (C18:3 and C22:6).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71195

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human fatty acid transport protein 2a/very long chain acyl-CoA synthetase 1 (FATP2a/Acsvl1) has a preference in mediating the channeling of exogenous n-3 fatty acids into phosphatidylinositol.
Melton EM, et al.
The Journal of Biological Chemistry, 286, 30670-30679 (2011)
Fatty acid transport protein-2 inhibitor Grassofermata/CB5 protects cells against lipid accumulation and toxicity.
Saini N, et al.
Biochemical and Biophysical Research Communications, 465, 534-541 (2015)
Emman Shubbar et al.
BMC cancer, 13, 1-1 (2013-01-04)
Breast cancer is a potentially fatal malignancy in females despite the improvement in therapeutic techniques. The identification of novel molecular signatures is needed for earlier detection, monitoring effects of treatment, and predicting prognosis. We have previously used microarray analysis to
Assignment of human fatty-acid-coenzyme A ligase, very long-chain 1 gene (FACVL1) to human chromosome band 15q21.2 by fluorescence in situ hybridization.
K Wakui et al.
Cytogenetics and cell genetics, 81(3-4), 292-293 (1998-09-08)
S J Steinberg et al.
Molecular genetics and metabolism, 68(1), 32-42 (1999-09-10)
Activation of fatty acids, catalyzed by acyl-coenzyme A (acyl-CoA) synthetases, is required for their subsequent metabolism. Peroxisomes and microsomes contain very-long-chain acyl-CoA synthetases (VLCSs) capable of activating fatty acids with a chain length of 22 or more carbons. Decreased peroxisomal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service