Skip to Content
Merck
All Photos(2)

Key Documents

HPA026088

Sigma-Aldrich

Anti-GPR61 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BALGR, Anti-G protein-coupled receptor 61

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

QFVCFFKPAPEEELRLPSREGSIEENFLQFLQGTGCPSESWVSRPLPSPKQEPPAVDFRIPGQIAEETSEFLEQQLTSDIIMSDSYLRPAASPRLES

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPR61(83873)

General description

G protein-coupled receptor 61 (GPR61) is expressed in the brain. The gene encoding it is localized on human chromosome 1.

Immunogen

G protein-coupled receptor 61 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

G protein-coupled receptor 61 (GPR61) may have a physiological role in the nervous system. It has been shown to associate with G-proteins. Studies have shown that GPR61 deficiency in mice causes obesity associated with hyperphagia, which indicates that it may be involved in the regulation of food-intake and body weight.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71175

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masaru Toyooka et al.
Journal of neuroscience research, 87(6), 1329-1333 (2008-11-26)
G-protein-coupled receptor 61 (GPR61) is an orphan receptor that is abundantly expressed in the brain, which suggests its involvement in various physiological functions in the central nervous system. It couples with Gs and shows constitutive activity. To investigate the role
Tom Walsh et al.
American journal of human genetics, 87(1), 90-94 (2010-07-07)
Massively parallel sequencing of targeted regions, exomes, and complete genomes has begun to dramatically increase the pace of discovery of genes responsible for human disorders. Here we describe how exome sequencing in conjunction with homozygosity mapping led to rapid identification
S Cikos et al.
Biochimica et biophysica acta, 1521(1-3), 66-72 (2001-11-03)
We report the cloning and sequence of a novel gene, BALGR, which is coding for a candidate G protein-coupled receptor (GPCR) that is distantly related to the histamine, adrenergic, serotonin and dopamine receptors. The coding region of the human BALGR
Hirohide Nambu et al.
Life sciences, 89(21-22), 765-772 (2011-10-06)
GPR61 is an orphan G protein-coupled receptor whose function remains unknown. The purpose of the present study is to elucidate the importance of GPR61 in metabolism by characterization of GPR61-deficient mice. Male GPR61-deficient mice were characterized regarding various metabolic parameters

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service