Skip to Content
Merck
All Photos(7)

Key Documents

HPA023149

Sigma-Aldrich

Anti-IDO1 antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Ido1 Antibody, Ido1 Antibody - Anti-IDO1 antibody produced in rabbit, Anti-IDO, Anti-Indoleamine 2,3-dioxygenase, Anti-Indoleamine-pyrrole 2,3-dioxygenase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€482.00

€482.00


Check Cart for Availability


Select a Size

Change View
100 μL
€482.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€482.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

LCSLESNPSVREFVLSKGDAGLREAYDACVKALVSLRSYHLQIVTKYILIPASQQPKENKTSEDPSKLEAKGTGGTDLMNFLK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IDO1(3620)

General description

IDO1 (indoleamine 2,3-dioxygenase 1) protein is present in the cytoplasm and exists as a monomer. It is mainly expressed in parenchymal tissues including, lungs, gut and the fetal-maternal unit during pregnancy. In addition, it can be seen in trophoblast, fibroblasts, epithelial and tumor cells, tumor-associated cells, macrophages, dendritic cells and microglial cells in the central nervous system.[1] The gene is mapped to human chromosome 8p11. It is also referred to as INDO (indoleamine-pyrrole 2,3-dioxygenase).

Immunogen

Indoleamine 2,3-dioxygenase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

IDO1 (indoleamine 2, 3-dioxygenase 1) is involved in the metabolism of tryptophan into kynurenine (Kyn) in extrahepatic tissues. It is needed for the first step in the Kyn pathway. IDO1 plays a crucial role in the innate host defenses by suppressing microbial growth. In addition, it enhances the immune tolerance by allowing maturation of naive T lymphocytes into T regulatory cells. It is upregulated in cancer and HIV (human immunodeficiency virus)-disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76238

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of IDO1 and PD-L2 in Patients with Benign Lymphadenopathies and Association with Autoimmune Diseases.
Abdulla, et al.
Biomolecules, 13 (2023)
Roberta Mancuso et al.
PloS one, 10(6), e0130715-e0130715 (2015-06-26)
Interferon gamma (IFN-γ) production induces the transcription of indoleamine 2,3 dioxygenase (IDO) resulting in the reduction of T-cell activation and proliferation through the depletion of tryptophan and the elicitation of Treg lymphocytes. IDO was shown to be involved in the
Iona Cheng et al.
Genes, chromosomes & cancer, 51(1), 66-76 (2011-10-04)
Detecting genomic alterations that result in more aggressive prostate cancer may improve clinical treatment and our understanding of the biology underlying this common but complex disease. To this end, we undertook a genome-wide copy number alterations (CNAs) study of clinicopathological
Guanyou Huang et al.
International journal of molecular sciences, 13(9), 10863-10879 (2012-10-31)
Indoleamine 2,3-dioxygenase (IDO) has been implicated in preventing the fetus from undergoing maternal T cell-mediated immune responses, yet the mechanism underlying these kinds of IDO-mediated immune responses has not been fully elucidated. Since the CD4 molecule plays a central role
Catharina Bartmann et al.
American journal of reproductive immunology (New York, N.Y. : 1989), 75(5), 539-556 (2016-02-04)
Human pregnancy needs a remarkable local immune tolerance toward the conceptus. Myeloid-derived suppressor cells (MDSC) are important players promoting cancer initiation and progression by suppressing T-cell functions and thus inducing immune tolerance. Therefore, MDSC were expected within decidua. Subpopulations of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service