Skip to Content
Merck
All Photos(8)

Documents

HPA022845

Sigma-Aldrich

Anti-NEFM antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-160 kDa neurofilament protein, Anti-NEF3 antibody produced in rabbit, Anti-NF-M, Anti-Neurofilament 3, Anti-Neurofilament medium polypeptide, Anti-Neurofilament triplet M protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

KEEEEKEVKEAPKEEKVEKKEEKPKDVPEKKKAESPVKEEAVAEVVTITKSVKVHLEKETKEEGKPLQQEKEKEKAGGEGGSEEEGSDKGAKG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEF3(4741)

General description

NEFM (neurofilament, medium polypeptide) is an abundant intermediate filament (IF) protein in post-mitotic neurons. It is mapped to human chromosome 8q21. It is abundantly present in the nervous system. In human, the protein has 13 carboxy-terminal domains with phosphorylation sites.

Immunogen

Neurofilament medium polypeptide recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NEFM (neurofilament, medium polypeptide) protein is involved in the axonal transport and neuronal cell apoptosis. Phosphorylation of NEFM allows the association between NEFM and microtubules. Deregulated phosphorylation of NEFM is associated with Alzheimer disease (AD). From gene mutation analysis, it has been predicted that NEFM may act as a susceptibility factor for the Parkinson′s disease (PD).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76206

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rejko Krüger et al.
Neuroscience letters, 351(2), 125-129 (2003-10-30)
Neurofilament M, a major component of Lewy bodies, represents an interesting candidate in the pathogenesis of Parkinson's disease (PD). We performed detailed mutation analyses of the NF-M gene in 322 familial and sporadic PD patients. Two polymorphisms (Ala475Thr and Gly697Arg)
Kathryn L Penney et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 24(1), 255-260 (2014-11-06)
Numerous germline genetic variants are associated with prostate cancer risk, but their biologic role is not well understood. One possibility is that these variants influence gene expression in prostate tissue. We therefore examined the association of prostate cancer risk variants
Parvathi Rudrabhatla et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 24(11), 4396-4407 (2010-07-14)
Aberrant hyperphosphorylation of neuronal cytoskeletal proteins is one of the major pathological hallmarks of neurodegenerative disorders such as Alzheimer disease (AD), amyotrophic lateral sclerosis (ALS), and Parkinson's disease (PD). Human NF-M/H display a large number of multiple KSP repeats in
Raul Perez-Olle et al.
Journal of neuropathology and experimental neurology, 63(7), 759-774 (2004-08-05)
The human neurofilament medium (hNFM) subunit is one of the 3 neurofilament (NF) polypeptides, which are the most abundant intermediate filament (IF) proteins in post-mitotic neurons. The formation of neurofilamentous aggregates is a pathological hallmark of many neurodegenerative diseases, including
Carmela Maniero et al.
Hypertension (Dallas, Tex. : 1979), 70(2), 357-364 (2017-06-07)
Heterogeneity among aldosterone-producing adenomas (APAs) has been highlighted by the discovery of somatic mutations. KCNJ5 mutations predominate in large zona fasciculata (ZF)-like APAs; mutations in CACNA1D, ATP1A1, ATP2B3, and CTNNB1 are more likely to be found in small zona glomerulosa

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service