Skip to Content
Merck
All Photos(3)

Key Documents

HPA021816

Sigma-Aldrich

Anti-NUP88 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-88 kDa nuclear pore complex protein, Anti-Nuclear pore complex protein Nup88, Anti-Nucleoporin Nup88

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

QLLTRNVVFGLGGELFLWDGEDSSFLVVRLRGPSGGGEEPALSQYQRLLCINPPLFEIYQVLLSPTQHHVALIGIKGLMVLELPKRWGKNSEFEGGKSTVNCSTTPVAERFFTSSTSLTLKHAAWYPSEILDPHVV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NUP88(4927)

General description

NUP88 (Nucleoporin 88kDa) is an 88kDa non-FG nucleoporin. It acts as a component of novel nuclear pore complex (NPC). It consists of a C-terminal coiled-coil domain and an interaction domain. It is localized in the cytoplasmic surface of nuclear pore complexes (NPCs). NUP88 consists of two domains, N-terminal and C-terminal domains and its N-terminal and C-terminal domains have been predicted to form β-propeller and a coiled-coil domain.

Immunogen

Nuclear pore complex protein Nup88 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NUP88 (Nucleoporin 88kDa) participates in trafficking between the nucleus and cytoplasm. Its N-terminal β-propeller domain interacts with the FG (phenylalanine-glycine) repeat domain of nucleoporin CAN/Nup214. Futher it binds directly to CRM-1/exportin-1, the receptor for export of most proteins from the nucleus. Nup88 also possesses the binding ability for the other FG nucleoporins Nup358 and Nup98. Overexpression of NUP88 has been observed in several types of malignancies such as colorectal cancer (CRC), breast cancer cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75556

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Fornerod et al.
The EMBO journal, 16(4), 807-816 (1997-02-17)
The oncogenic nucleoporin CAN/Nup214 is essential in vertebrate cells. Its depletion results in defective nuclear protein import, inhibition of messenger RNA export and cell cycle arrest. We recently found that CAN associates with proteins of 88 and 112 kDa, which
Chieko Hashizume et al.
Molecular cancer, 9, 119-119 (2010-05-26)
Nuclear pore complexes are massive multiprotein channels responsible for traffic between the nucleus and cytoplasm, and are composed of approximately 30 proteins, termed nucleoporins (Nup). Our recent studies indicated that the nucleoporins Rae1 and Tpr play critical roles in maintaining

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service