Skip to Content
Merck
All Photos(7)

Key Documents

HPA021545

Sigma-Aldrich

Anti-TPK1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-PP20, Anti-Placental protein 20, Anti-Thiamin pyrophosphokinase 1, Anti-Thiamine pyrophosphokinase 1, Anti-hTPK1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50
western blot: 0.04-0.4 μg/mL

immunogen sequence

HRLHVDTGMEGDWCGLIPVGQPCMQVTTTGLKWNLTNDVLAFGTLVSTSNTYDGSGVVTVETDHPL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TPK1(27010)

General description

TPK1 (thiamine pyrophosphokinase 1) protein is encoded by TPK1 gene localized to human chromosome 7q34-36. The gene comprises of at least nine exons and spans ~420kb. TPK1 enzyme is highly expressed in maternal, placental and fetal tissues. The human TPK1 mRNA is expressed relatively high in kidney, small intestine and testis with lesser expression in brain, liver, placenta and spleen.

Immunogen

Thiamin pyrophosphokinase 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TPK1 (thiamine pyrophosphokinase 1) is a cellular enzyme that catalyzes the conversion of thiamine to thiamine pyrophosphate (TPP). TPK1 enzyme plays a vital role in controlling birth weight. Mutation of this gene causes thiamine metabolism dysfunction called episodic encephalopathy .

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75128

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siddharth Banka et al.
Molecular genetics and metabolism, 113(4), 301-306 (2014-12-03)
Thiamine pyrophosphokinase (TPK) produces thiamine pyrophosphate, a cofactor for a number of enzymes, including pyruvate dehydrogenase and 2-ketoglutarate dehydrogenase. Episodic encephalopathy type thiamine metabolism dysfunction (OMIM 614458) due to TPK1 mutations is a recently described rare disorder. The mechanism of
D Fradin et al.
Annals of human genetics, 71(Pt 5), 578-585 (2007-02-14)
Extreme variations in birth weight increase immediate postnatal mortality and morbidity, and are also associated with the predisposition to metabolic diseases in late adulthood. Birth weight in humans is influenced by yet unknown genetic factors. Since the 7q34-q35 region showed
R Zhao et al.
Biochimica et biophysica acta, 1517(2), 320-322 (2001-05-09)
Thiamin pyrophosphokinase (TPK, EC 2.7.6.2) catalyses phosphorylation of thiamin to thiamin pyrophosphate, an active enzyme cofactor. Here we describe the cloning of complete human TPK1 cDNA from an adult liver library. Human TPK1 is 89% identical to murine TPK1 at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service