Skip to Content
Merck
All Photos(3)

Key Documents

HPA021407

Sigma-Aldrich

Anti-ARHGDIA antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Rho GDI 1, Anti-Rho GDP-dissociation inhibitor 1, Anti-Rho-GDI alpha

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

MAEQEPTAEQLAQIAAENEEDEHSVNYKPPAQKSIQEIQELDKDDESLRKYKEALLGRVAVSAD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARHGDIA(396)

Related Categories

General description

The gene ARHGDIA (rho GDP-dissociation inhibitor 1) is mapped to human chromosome 17q25.3. It belongs to the RhoGDI (rho GDP-dissociation inhibitor) family of proteins. ARHGDIA is widely expressed in human tissues.

Immunogen

Rho GDP-dissociation inhibitor 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ARHGDIA (rho GDP-dissociation inhibitor 1) is mainly responsible for holding Rho (ras homolog) proteins in an inactive state in the cytoplasm, thereby protecting them from degradation. It is down-regulated in hepatocellular carcinoma (HCC). Absence of ARHGDIA helps in hepatic tumorigenesis.[1] ARHGDIA is also associated with nephrotic syndrome.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75670

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Indra Rani Gupta et al.
Journal of medical genetics, 50(5), 330-338 (2013-02-26)
Congenital nephrotic syndrome arises from a defect in the glomerular filtration barrier that permits the unrestricted passage of protein across the barrier, resulting in proteinuria, hypoalbuminaemia, and severe oedema. While most cases are due to mutations in one of five
Zipeng Cao et al.
Molecular oncology, 8(2), 285-296 (2013-12-18)
Selective activation of Rho GTPase cascade requires the release of Rho from RhoGDI (GDP-dissociation inhibitors) complexes. Our previous studies identified RhoGDIα SUMOylation at Lys-138 and its function in the regulation of cancer cell invasion. In the current study, we demonstrate
Raquel Bermudo et al.
BMC cancer, 8, 315-315 (2008-11-01)
Transcriptional profiling of prostate cancer (PC) has unveiled new markers of neoplasia and allowed insights into mechanisms underlying this disease. Genomewide analyses have also identified new chromosomal abnormalities associated with PC. The combination of both classes of data for the
Etienne Boulter et al.
Nature cell biology, 12(5), 477-483 (2010-04-20)
At steady state, most Rho GTPases are bound in the cytosol to Rho guanine nucleotide dissociation inhibitors (RhoGDIs). RhoGDIs have generally been considered to hold Rho proteins passively in an inactive state within the cytoplasm. Here we describe an evolutionarily
Lei Liang et al.
International journal of oncology, 45(2), 659-666 (2014-05-27)
Rho GTPases control a wide range of cellular processes and contribute to tumor invasion and metastasis. As a regulator of Rho activity, ARHGDIA is aberrantly expressed in several types of tumors and plays different roles in the tumor process. To

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service