Skip to Content
Merck
All Photos(3)

Key Documents

HPA021403

Sigma-Aldrich

Anti-LRSAM1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-E3 ubiquitin-protein ligase LRSAM1, Anti-Leucine-rich repeat and sterile alpha motif-containing protein 1, Anti-Tsg101-associated ligase, Anti-hTAL

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20-1:50
western blot: 0.04-0.4 μg/mL

immunogen sequence

LENERIRMEQLMSITQEETESLRRRDVASAMQQMLTESCKNRLIQMAYESQRQNLVQQACSSMAEMDERFQQILSWQQMDQNKAISQI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LRSAM1(90678)

General description

The gene LRSAM1 (leucine-rich repeat and sterile α motif-containing protein 1) is mapped to human chromosome 9q33.3. The protein is predicted to contain a leucine-rich repeat (LRR), an ezrin-radixin-moezin (ERM) domain, a coiled-coil (CC) region, a SAM (sterile α motif) domain and a carboxyl-terminal C3HC4-type RING (really interesting new gene) finger domain.

Immunogen

E3 ubiquitin-protein ligase LRSAM1 recombinant protein epitope signature tag (PrEST)

Application

Anti-LRSAM1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

LRSAM1 (leucine-rich repeat and sterile α motif-containing protein 1) is an E3 ligase. It binds with and ubiquitinates Tsg101 (tumor susceptibility gene 101), inactivating Tsg101-mediated sorting of endocytic (epidermal growth factor receptors) and exocytic (viral proteins) cargoes. It also protects cytoplasm from invasive pathogens by participating in ubiquitination associated with intracellular bacteria. Mutations in LRSAM1 are linked with Charcot-Marie-Tooth disease.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75320

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thalia Antoniadi et al.
BMC medical genetics, 16, 84-84 (2015-09-24)
Inherited peripheral neuropathy (IPN) is a clinically and genetically heterogeneous group of disorders with more than 90 genes associated with the different subtypes. Sequential gene screening is gradually being replaced by next generation sequencing (NGS) applications. We designed and validated
Laurent P Bogdanik et al.
Disease models & mechanisms, 6(3), 780-792 (2013-03-23)
Charcot-Marie-Tooth disease (CMT) is a clinically and genetically heterogeneous condition characterized by peripheral axon degeneration with subsequent motor and sensory deficits. Several CMT gene products function in endosomal sorting and trafficking to the lysosome, suggesting that defects in this cellular
Duane L Guernsey et al.
PLoS genetics, 6(8), doi:10-doi:10 (2010-09-25)
Charcot-Marie-Tooth disease (CMT) represents a family of related sensorimotor neuropathies. We studied a large family from a rural eastern Canadian community, with multiple individuals suffering from a condition clinically most similar to autosomal recessive axonal CMT, or AR-CMT2. Homozygosity mapping
Ido Amit et al.
Genes & development, 18(14), 1737-1752 (2004-07-17)
The tumor suppressor gene 101 (tsg101) regulates vesicular trafficking processes in yeast and mammals. We report a novel protein, Tal (Tsg101-associated ligase), whose RING finger is necessary for multiple monoubiquitylation of Tsg101. Bivalent binding of Tsg101 to a tandem tetrapeptide
Alan Huett et al.
Cell host & microbe, 12(6), 778-790 (2012-12-19)
Several species of pathogenic bacteria replicate within an intracellular vacuolar niche. Bacteria that escape into the cytosol are captured by the autophagic pathway and targeted for lysosomal degradation, representing a defense against bacterial exploitation of the host cytosol. Autophagic capture of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service