Skip to Content
Merck
All Photos(6)

Key Documents

HPA021154

Sigma-Aldrich

Anti-EBAG9 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Cancer-associated surface antigen RCAS1, Anti-Estrogen receptor-binding fragment-associated gene 9 protein, Anti-Receptor-binding cancer antigen expressed on SiSo cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

DGSTGFSSRLAATQDLPFIHQSSELGDLDTWQENTNAWEEEEDAAWQAEEVLRQQKLADREKRAAEQQRKKMEKEAQRLMKKEQNKIGVKL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EBAG9(9166)

General description

The gene EBAG9 (estrogen receptor-binding fragment-associated gene 9) is mapped to human chromosome 8q23. It is a type II transmembrane protein and is ubiquitously expressed. EBAG9 localizes at the Golgi complex and also exists in soluble form. It is commonly referred as RCAS1 (receptor-binding cancer antigen expressed on SiSo cells).

Immunogen

Receptor-binding cancer antigen expressed on SiSo cells recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

EBAG9 (estrogen receptor-binding fragment-associated gene 9) is up-regulated in many cancers, including uterine, ovarian, lung and breast cancer cells. It is suggested to mediate growth arrest and apoptosis in activated immune cells, and thereby helps tumor cells to escape host immune system.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73996

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Tsuneizumi et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 7(11), 3526-3532 (2001-11-14)
EBAG9, an estrogen-responsive gene located at 8q23 was identified in an effort to clone CpG-binding sites. Its product was later found to be identical to RCAS1, a cancer cell-surface antigen implicated in immune escape. We determined the sequence of the
Joel Rousseau et al.
Biochemical and biophysical research communications, 293(5), 1544-1549 (2002-06-11)
RCAS1/EBAG9 (receptor-binding cancer antigen expressed on SiSo cells/ estrogen receptor-binding fragment-associated gene 9), an estrogen-transcribed protein, has been shown to be expressed in a wide variety of cancers, including uterine, ovarian, and lung cancer cells. Soluble and membranous RCAS1 proteins
Satoshi Oizumi et al.
Oncology, 62(4), 333-339 (2002-07-26)
RCAS1 (receptor-binding cancer antigen expressed on SiSo cells) is a new tumor-associated antigen recognized by 22-1-1 monoclonal antibody. RCAS1 expressed on human cancer cells acts as a ligand for a putative receptor present on peripheral lymphocytes. RCAS1 has been shown
Arne Engelsberg et al.
The Journal of biological chemistry, 278(25), 22998-23007 (2003-04-04)
Tumor immunology has received a large impetus from the identification of tumor-associated antigens. Among them, a monoclonal antibody, 22.1.1, was instrumental in defining a novel tumor-associated antigen that was termed "receptor binding cancer antigen expressed on SiSo cells" (RCAS1). RCAS1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service