Skip to Content
Merck
All Photos(8)

Key Documents

HPA021062

Sigma-Aldrich

Anti-NOP58 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-NOP58, Anti-Nucleolar protein 5, Anti-Nucleolar protein NOP5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LDKELNNYIMRCREWYGWHFPELGKIISDNLTYCKCLQKVGDRKNYASAKLSELLPEEVEAEVKAAAEISMGTEVSEEDICNILHLCTQVIEISEYRTQLYEYLQNRMMAIAPNVTVMVGELVGARLIAHAGSLL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NOP58(51602)

General description

The gene nucleolar protein 58 (NOP58) is mapped to human chromosome 2q33.1. The protein is present in nucleoplasm and nucleolus.[1]

Immunogen

Nucleolar protein 5 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Nucleolar protein 58 (NOP58) is a component of box C/D small nucleolar ribonucleoprotein complexes (snoRNPs). SUMOylation of NOP58 is essential for binding to small nucleolar ribonucleoprotein complex. NOP58 is required for binding and retention of box C/D snoRNAs, such as U3, in the nucleolus.[1] RNAi mediated silencing of NOP58 results in strong reduction in U3 snoRNA levels. NOP58 associates with NOP56, TIP48 (TATA box-binding protein-interacting protein), TIP49, BCD1 (B-cell-derived protein 1), NOP17, NUFIP (nuclear FMRP-interacting protein), and TAF9 (Transcription initiation factor TFIID subunit 9). NOP58 also interacts with the nuclear import receptor subunit, importin α.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73943

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S K Lyman et al.
RNA (New York, N.Y.), 5(12), 1597-1604 (1999-12-22)
We have identified an apparent human homolog of the yeast Nop5/Nop58 protein. hNop5/Nop58 codes for a protein of predicted molecular weight 59.6 kDa and is 46.8% identical to Saccharomyces cerevisiae Nop5/Nop58. Immunofluorescent staining with antibodies against hNop5/Nop58 indicate that it
Belinda J Westman et al.
Molecular cell, 39(4), 618-631 (2010-08-28)
Posttranslational SUMO modification is an important mechanism of regulating protein function, especially in the cell nucleus. The nucleolus is the subnuclear organelle responsible for rRNA synthesis, processing, and assembly of the large and small ribosome subunits. Here, we have used
Kenneth Scott McKeegan et al.
Molecular and cellular biology, 27(19), 6782-6793 (2007-07-20)
The box C/D small nucleolar RNPs (snoRNPs) are essential for the processing and modification of rRNA. The core box C/D proteins are restructured during human U3 box C/D snoRNP biogenesis; however, the molecular basis of this is unclear. Here we
Ilaria Del Giudice et al.
American journal of hematology, 89(1), 74-82 (2013-09-14)
Chronic lymphocytic leukemia (CLL) with stereotyped B-cell receptor (BCR) belonging to subset #1 (IGHV1-5-7/ IGKV1-39) display a poor outcome. To characterize their genetic and genomic features and BCR function, we selected 20 subset #1 CLL from a series of 579

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service