Skip to Content
Merck
All Photos(2)

Key Documents

HPA021046

Sigma-Aldrich

Anti-UBR4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-600 kDa retinoblastoma protein-associated factor, Anti-E3 ubiquitin-protein ligase UBR4, Anti-N-recognin-4, Anti-RBAF600, Anti-Retinoblastoma-associated factor of 600 kDa, Anti-Zinc finger UBR1-type protein 1, Anti-p600

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Check Cart for Availability


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

VEFGGNDLLQVYNAQQIKHRLNSTGMYVANTKPGGFTIEISNNNSTMVMTGMRIQIGTQAIERAPSYIEIFGRTMQLNLSRSRWFDFPFTREEALQADKKLNLFIGASVDPAGVTMIDAVKIYGKTKEQF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBR4(23352)

General description

The gene UBR4 (ubiquitin protein ligase E3 component n-recognin 4) is mapped to human chromosome 1p36.13. The protein localizes in the cytoplasm and the nucleus. UBR4 is also referred to as p600.

Immunogen

E3 ubiquitin-protein ligase UBR4 recombinant protein epitope signature tag (PrEST)

Application

Anti-UBR4 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

UBR4 (ubiquitin protein ligase E3 component n-recognin 4) interacts with retinoblastoma protein in the nucleus and functions as a chromatin scaffold. In the cytoplasm, UBR4 regulates cytoskeletal dynamics and membrane morphogenesis. It is important for formation of integrin-mediated ruffled membrane and cell survival. UBR4 forms an E2-E3 complex with RAD6 (ubiquitin-conjugating enzyme E2) and KCMF1 (potassium channel modulatory factor 1). The complex regulates lysosome-mediated degradation of N-end rule substrates. UBR4 associates with human papillomavirus type 16 (HPV-16) E7 oncoprotein and participates in anchorage-independent growth in cancer cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72607

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kari Parsons et al.
Cellular and molecular life sciences : CMLS, 72(6), 1149-1160 (2014-11-27)
A decade ago, the large 600 kDa mammalian protein p600 (also known as UBR4) was discovered as a multifunctional protein with roles in anoikis, viral transformation and protein degradation. Recently, p600 has emerged as a critical protein in the mammalian brain
Jenny H Hong et al.
Molecular & cellular proteomics : MCP, 14(3), 674-685 (2015-01-15)
RAD6 is a ubiquitin E2 protein with roles in a number of different biological processes. Here, using affinity purification coupled with mass spectrometry, we identify a number of new RAD6 binding partners, including the poorly characterized ubiquitin E3 ligases KCMF1
Correction: Novel Genetic Locus Implicated for HIV-1 Acquisition with Putative Regulatory Links to HIV Replication and Infectivity: A Genome-Wide Association Study.
Eric O Johnson et al.
PloS one, 10(5), e0129671-e0129671 (2015-05-30)
Yoshihiro Nakatani et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(42), 15093-15098 (2005-10-11)
In this article, we identify and characterize p600, a unique 600-kDa retinoblastoma protein- and calmodulin-binding protein. In the nucleus, p600 and retinoblastoma protein seem to act as a chromatin scaffold. In the cytoplasm, p600 and clathrin form a meshwork structure
Liam C Hunt et al.
Cell reports, 28(5), 1268-1281 (2019-08-01)
Skeletal muscle cell (myofiber) atrophy is a detrimental component of aging and cancer that primarily results from muscle protein degradation via the proteasome and ubiquitin ligases. Transcriptional upregulation of some ubiquitin ligases contributes to myofiber atrophy, but little is known

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service