Skip to Content
Merck
All Photos(8)

Key Documents

HPA021011

Sigma-Aldrich

Anti-FGL2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Fibrinogen-like protein 2, Anti-Fibroleukin, Anti-pT49

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20-1:50

immunogen sequence

RNGLLLPSTGAPGEVGDNRVRELESEVNKLSSELKNAKEEINVLHGRLEKLNLVNMNNIENYVDSKVANLTFVVNSLDGKCSKCP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGL2(10875)

General description

The gene FGL2 (fibrinogen-like protein 2) is mapped to human chromosome 7q11.23. It is a transmembrane protein. FGL2 is present on cell surface of endothelial cells and activated macrophages. Additionally, it can be present in secreted form.

Immunogen

Fibroleukin Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FGL2 (fibrinogen-like protein 2) behaves as an immune-suppressive modulator and a prothrombinase. It cleaves prothrombin to thrombin and exerts coagulant activity. In addition, it can induce PD1 (programmed cell death protein 1) and CD39 levels, promote number of MDSCs (myeloid-derived suppressor cells) and CD39+ regulatory T cells, and increase occurrence of tumor-supportive M2 macrophages. FGL2 is up-regulated in many cancers. FGL2 is up-regulated in humans suffering from inflammatory bowel disease. The serum levels of FGL2 increase in humans with systemic sclerosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74958

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esther Rabizadeh et al.
PloS one, 9(10), e109648-e109648 (2014-10-11)
Fibrinogen-like protein 2, FGL-2, was reported to be overexpressed in various cancer tissues, where it acts as a transmembrane prothrombinase. This study aims to determine the prothrombinase activity of FGL-2 in peripheral blood mononuclear cells (PBMC) of patients with B-cell
Koichi Yanaba et al.
Clinical rheumatology, 32(1), 43-47 (2012-09-18)
Fibrinogen-like protein 2 (FGL2), a member of the fibrinogen-related superfamily of proteins, is expressed on the surface of macrophages, T cells, and endothelial cells and directly cleaves prothrombin to thrombin. The aim of this study is to determine the serum
Jun Yan et al.
Journal of the National Cancer Institute, 107(8), doi:10-doi:10 (2015-05-15)
Fibrinogen-like protein 2 (FGL2) may promote glioblastoma multiforme (GBM) cancer development by inducing multiple immune-suppression mechanisms. The biological significance of FGL2 expression was assessed using the The Cancer Genome Atlast (TCGA) glioma database and tumor lysates analysis. The therapeutic effects
S Yuwaraj et al.
Genomics, 71(3), 330-338 (2001-02-15)
For diseases in which thrombosis plays a pivotal role, such as virus-induced fulminant hepatitis, fetal loss syndrome, and xenograft rejection, the major procoagulant has remained elusive. Here we describe the isolation and functional expression of a distinct human prothrombinase, termed
Xiuli Dong et al.
Digestive diseases and sciences, 59(4), 769-777 (2013-11-30)
Fibrinogen-like protein 2 (FGL2), a new member of the fibrinogen-like family, has recently been identified as a novel immunosuppressive molecule. The purpose of this work was to investigate intestinal and peripheral expression of FGL2 in patients with inflammatory bowel disease

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service