Skip to Content
Merck
All Photos(3)

Key Documents

HPA020952

Sigma-Aldrich

Anti-PHPT1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-14 kDa phosphohistidine phosphatase, Anti-Phosphohistidine phosphatase 1, Anti-Protein janus-A homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

DLALIPDVDIDSDGVFKYVLIRVHSAPRSGAPAAESKEIVRGYKWAEYHADIYDKVSGDMQKQGCDCECLGGGRISHQSQDKKIHVYGYSMAYGPAQHAISTEKIKAKYPDYE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PHPT1(29085)

General description

The gene PHPT1 (phosphohistidine phosphatase 1) is mapped to human chromosome 9q34.3. It is widely expressed in human tissues.

Immunogen

14 kDa phosphohistidine phosphatase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PHPT1 (phosphohistidine phosphatase 1) is mainly responsible for dephosphorylation of phosphohistidine in target proteins. PHPT1 substrates include epithelial Ca2+ channel TRPV5 (transient receptor potential cation channel subfamily V member 5), ATP (adenosine triphosphate)-citrate lyase and β-subunit of G protein. In lung cancer, PHPT1 is linked with carcinogenesis and metastasis. There are reports of PHPT1 involvement in dephosphorylation of phospholysine in chemically phosphorylated histone H1.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73827

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pia Ek et al.
Upsala journal of medical sciences, 120(1), 20-27 (2015-01-13)
Phosphohistidine phosphatase 1 (PHPT1), also named protein histidine phosphatase (PHP), is a eukaryotic enzyme dephosphorylating proteins and peptides that are phosphorylated on a histidine residue. A preliminary finding that histone H1, which lacks histidine, was phosphorylated by phosphoramidate and dephosphorylated
Xinjiang Cai et al.
Molecular biology of the cell, 25(8), 1244-1250 (2014-02-14)
The kidney, together with bone and intestine, plays a crucial role in maintaining whole-body calcium (Ca(2+)) homoeostasis, which is primarily mediated by altering the reabsorption of Ca(2+) filtered by the glomerulus. The transient receptor potential-vanilloid-5 (TRPV5) channel protein forms a
An-Jian Xu et al.
Chinese medical journal, 123(22), 3247-3251 (2010-12-18)
In our previous studies, we found the expression of 14-kD phosphohistidine phosphatase (PHPT1) was associated with lung cancer cells migration and invasion, and PHPT1 mRNA expression level in lung cancer tissues clinically correlated with lymph node metastasis. In the present
Raviteja Inturi et al.
The international journal of biochemistry & cell biology, 57, 69-75 (2014-12-03)
Regulation of protein activity by phosphorylation is central in many cellular processes. Phosphorylation of serine, threonine and tyrosine residues is well documented and studied. In addition, other amino acids, like histidine can be phosphorylated, but neither the mechanism nor the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service