Skip to Content
Merck
All Photos(6)

Key Documents

HPA020949

Sigma-Aldrich

Anti-ZMYND8 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CTCL tumor antigen se14-3, Anti-Cutaneous T-cell lymphoma-associated antigen se14-3, Anti-Protein kinase C-binding protein 1, Anti-Rack7, Anti-Zinc finger MYND domain-containing protein 8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Check Cart for Availability


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

TTKTDKTSTTGSILNLNLDRSKAEMDLKELSESVQQQSTPVPLISPKRQIRSRFQLNLDKTIESCKAQLGINEISEDVYTAVEHSDSEDSEKSDSSDSEYISDDEQKSKN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZMYND8(23613)

General description

The gene ZMYND8 (zinc finger MYND domain-containing protein 8) is mapped to human chromosome 20q13.12. The protein has a plant homeodomain, a bromodomain, a PWWP (pro-trp-trp-pro) chromatin-binding domain and a protein-protein interaction MYND (myeloid, Nervy, and DEAF-1) domain.

Immunogen

Protein kinase C-binding protein 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-ZMYND8 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting.[1] To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige. In addition, the antibody has been used for co-immunoprecipitation.[1]

Biochem/physiol Actions

ZMYND8 (zinc finger MYND domain-containing protein 8) is involved in DNA damage response. It brings the nucleosome remodeling and histone deacetylation complex to damaged chromatin, and promotes homologous recombination. ZMYND8 has been shown as a cutaneous T-cell lymphoma-associated antigen. In acute erythroid leukaemia, ZMYND8-RELA (v-rel avian reticuloendotheliosis viral oncogene homolog A) fusion gene increases leukaemogenically. ZMYND8 induces expression of vascular endothelial growth factor and thereby mediates tumor angiogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76008

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fade Gong et al.
Genes & development, 29(2), 197-211 (2015-01-17)
How chromatin shapes pathways that promote genome-epigenome integrity in response to DNA damage is an issue of crucial importance. We report that human bromodomain (BRD)-containing proteins, the primary "readers" of acetylated chromatin, are vital for the DNA damage response (DDR).
Ioannis Panagopoulos et al.
PloS one, 8(5), e63663-e63663 (2013-05-15)
Acute erythroid leukemia was diagnosed in a 4-month-old boy. Cytogenetic analysis of bone marrow (BM) cells showed a t(11;20)(p11;q11) translocation. RNA extracted from the BM was sequenced and analyzed for fusion transcripts using the software FusionMap. A ZMYND8-RELA fusion was
Michela Serresi et al.
Science advances, 7(9) (2021-02-26)
Epithelial-mesenchymal transition (EMT) is a developmental process hijacked by cancer cells to modulate proliferation, migration, and stress response. Whereas kinase signaling is believed to be an EMT driver, the molecular mechanisms underlying epithelial-mesenchymal interconversion are incompletely understood. Here, we show
Pan Jia et al.
Nature communications, 12(1), 6535-6535 (2021-11-13)
Super-enhancers (SEs) govern macrophage polarization and function. However, the mechanism underlying the signal-dependent latent SEs remodeling in macrophages remains largely undefined. Here we show that the epigenetic reader ZMYND8 forms liquid compartments with NF-κB/p65 to silence latent SEs and restrict
Verónica Delgado-Benito et al.
Molecular cell, 72(4), 636-649 (2018-10-09)
Class switch recombination (CSR) is a DNA recombination reaction that diversifies the effector component of antibody responses. CSR is initiated by activation-induced cytidine deaminase (AID), which targets transcriptionally active immunoglobulin heavy chain (Igh) switch donor and acceptor DNA. The 3'

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service