Skip to Content
Merck
All Photos(12)

Key Documents

HPA020912

Sigma-Aldrich

Anti-AMACR antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-α-methylacyl-CoA racemase, Anti-RACE

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

APFYTTYRTADGEFMAVGAIEPQFYELLIKGLGLKSDELPNQMSMDDWPEMKKKFADVFAKKTKAEWCQIFDGTDACVTPVLTF

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AMACR(23600)

General description

The gene AMACR (α-methylacyl-CoA racemase) is mapped to human chromosome 5p13.3.[1] The protein localizes in the mitochondria and peroxisomes.

Immunogen

alpha-methylacyl-CoA racemase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

AMACR (α-methylacyl-CoA racemase) is responsible for β-oxidation of dietary branched-chain fatty acids and synthesis of bile acid.[2] Mode of action involves interconversion of (R)- and (S)-2-methyl branched-chain fatty acyl-CoA fragments.[2] Mutations in AMACR are linked with adult-onset sensory motor neuropathy. It is up-regulated in myxofibrosarcomas, prostate cancer and nasopharyngeal carcinoma.[2]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74751

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying-En Lee et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(8), 7983-7991 (2014-05-17)
The molecular prognostic adjunct in patients with nasopharyngeal carcinomas (NPCs) still remains obscured. Through data mining from published transcriptomic database, alpha-methylacyl-CoA racemase (AMACR) was first identified as a differentially upregulated gene in NPC tissues, which is a key enzyme for
Bo Xu et al.
Journal of clinical pathology, 67(11), 974-979 (2014-08-06)
Hepatocellular carcinoma (HCC) is one of the most common malignancies worldwide, and it is still lacking effective prognostic biomarkers so far. Previous results of the iTRAQ-based quantitative proteomics study (iTRAQ-2DLC-MS/MS) have shown that α-methylacyl-CoA racemase (AMACR) might be a promising
Chien-Feng Li et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(23), 6141-6152 (2014-11-12)
Myxofibrosarcomas frequently display arm-level gains on 5p. We characterized the pathogenetic and therapeutic relevance of the α-methylacyl coenzyme A racemase (AMACR) at 5p13.3. AMACR mRNA expression in myxofibrosarcomas was analyzed using the public transcriptome and laser-microdissected sarcoma cells. We performed
Mariya Shapovalova et al.
Oncotarget, 9(94), 36693-36704 (2019-01-08)
The metabolic protein alpha-methylacyl-CoA racemase (AMACR) is significantly overexpressed in prostate cancer compared to the normal prostate and other non-malignant tissue. Though an attractive target, there are no reports in the literature on leveraging the expression of AMACR for the
S Ferdinandusse et al.
Nature genetics, 24(2), 188-191 (2000-02-02)
Sensory motor neuropathy is associated with various inherited disorders including Charcot-Marie-Tooth disease, X-linked adrenoleukodystrophy/adrenomyeloneuropathy and Refsum disease. In the latter two, the neuropathy is thought to result from the accumulation of specific fatty acids. We describe here three patients with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service