Skip to Content
Merck
All Photos(7)

Key Documents

HPA020376

Sigma-Aldrich

Anti-PSPH antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-L-3-phosphoserine phosphatase, Anti-O-phosphoserine phosphohydrolase, Anti-PSP, Anti-PSPase, Anti-Phosphoserine phosphatase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Estimated to ship on05 April 2025



Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Estimated to ship on05 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

AVCFDVDSTVIREEGIDELAKICGVEDAVSEMTRRAMGGAVPFKAALTERLALIQPSREQVQRLIAEQPPHLTPGIRELVSRLQERNVQVFLISG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PSPH(5723)

General description

Phosphoserine phosphatase (PSPH) is an Mg2+-dependent enzyme having a molecular mass of 25kDa. It belongs to the haloacid dehalogenase (HAD) superfamily and possesses 225 amino acids. PSPH is present in endosomes and has an amino terminal motif required for its activity. The gene encoding the enzyme is localized on human chromosome 7.

Immunogen

Phosphoserine phosphatase recombinant protein epitope signature tag (PrEST)

Application

Anti-PSPH antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Phosphoserine phosphatase (PSPH) takes part in the hydrolysis of L-phosphoserine and the formation of L-serine. Deficiency of the enzyme has been associated with Williams syndrome.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75004

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Kitazawa et al.
Oncotarget, 8(17), 28922-28938 (2017-04-21)
Mutations in succinate dehydrogenase B (SDHB) gene are frequently observed in several tumors and associated with poor prognosis in these tumors. Therefore, drugs effective for SDHB-deficient tumors could fulfill an unmet medical need. In addition, such drugs would have an
Kuniaki Sato et al.
Anticancer research, 37(5), 2365-2371 (2017-05-10)
Amplification of chromosome 7p (Ch.7p) is common in colorectal cancer (CRC). The aim of this study was to identify potential driver genes on Ch.7p that are overexpressed due to DNA copy number amplification and determine their clinical significance in CRC.
Michael A Bachelor et al.
Journal of dermatological science, 63(3), 164-172 (2011-07-06)
L-3-Phosphoserine phosphatase (PSPH) is a highly conserved and widely expressed member of the haloacid dehalogenase superfamily and the rate-limiting enzyme in l-serine biosynthesis. We previously found Psph expression to be uniquely upregulated in a α6β4 integrin transgenic mouse model that
Yves Peeraer et al.
European journal of biochemistry, 271(16), 3421-3427 (2004-08-05)
The structure of the Mg(2+)-dependent enzyme human phosphoserine phosphatase (HPSP) was exploited to examine the structural and functional role of the divalent cation in the active site of phosphatases. Most interesting is the biochemical observation that a Ca(2+) ion inhibits
Maria Veiga-da-Cunha et al.
European journal of human genetics : EJHG, 12(2), 163-166 (2003-12-16)
We report the identification of the mutations in the only known case of L-3-phosphoserine phosphatase deficiency, a recessively inherited condition. The two mutations correspond to the replacement of the semiconserved Asp32 residue by an asparagine and of the extremely conserved

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service