Skip to Content
Merck
All Photos(2)

Key Documents

HPA020057

Sigma-Aldrich

Anti-AIMP2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-JTV1 antibody produced in rabbit, Anti-Multisynthetase complex auxiliary component p38, Anti-Protein JTV-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

SVLGKDYGALKDIVINANPASPPLSLLVLHRLLCEHFRVLSTVHTHSSVKSVPENLLKCFGEQNKKQPRQDYQLGFTLIWKNVPKTQ

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... JTV1(7965)

General description

Aminoacyl tRNA synthetase complex interacting multifunctional protein 2 (AIMP2) is localized to the cytosol. It is part of the multi-tRNA synthetase complex. The AIMP2 gene is located on human chromosome 7.

Immunogen

Multisynthetase complex auxiliary component p38 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Aminoacyl tRNA synthetase complex interacting multifunctional protein 2 (AIMP2) is a scaffold protein which functions as a tumor suppressor. It stimulates the ubiquitin-mediated degradation of FUSE-binding protein (FUBP1), which is a transcription activator. AIMP2 activates tumor suppressor p53 and stimulates apoptosis. The assembly of macromolecular tRNA synthetase complex depends on AIMP2.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74923

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cancer-associated splicing variant of tumor suppressor AIMP2/p38: pathological implication in tumorigenesis.
Choi JW
PLoS Genetics, 7(3), e1001351-e1001351 (2011)
You-Sub Won et al.
Journal of biotechnology, 158(1-2), 44-49 (2012-01-31)
AIMP2/p38 is a scaffolding protein critical for the assembly of the macromolecular tRNA synthetase complex. Moreover, AIMP2 harbors anti-proliferative activity and promotes cell death as a proapototic factor. A splicing variant of AIMP2 lacking exon 2 (AIMP2-DX2) is specifically generated
Dae Gyu Kim et al.
Cancer research, 76(11), 3422-3436 (2016-05-20)
AIMP2/p38 is a multifunctional tumor suppressor that normally resides in the cytosol as a scaffold protein of the multi-tRNA synthetase complex (MSC). One of the tumor-suppressive functions of AIMP2 is to facilitate ubiquitin-mediated degradation of FUSE-binding protein (FBP, FUBP1), a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service