Skip to Content
Merck
All Photos(10)

Key Documents

HPA019907

Sigma-Aldrich

Anti-CCAR2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-DBC-1, Anti-DBC1, Anti-KIAA1967, Anti-NET35

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

MLLSLPEKVVSPPEPEKEEAAKEEATKEEEAIKEEVVKEPKDEAQNEGPATESEAPLKEDGLLPKPLSSGGEEEEKPRGEASEDLCEMALD

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

KIAA1967 is a large multi-domain containing nuclear protein mapped on chromosomal location 8p22. It consists of a NL (nuclear localization) motif, a CC (coiled-coil) domain responsible for the protein-protein interaction, an inactive EF-hand for Ca2+ binding, a Nudix domain, a RNA-binding domain and an LZ (leucine zipper) motif.

Immunogen

Protein KIAA1967 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

KIAA1967 is associated with several cellular processes such as regulation of metabolism, aging and tumor biology. It plays a vital role in the cancer pathogenesis by participating in apoptosis, nuclear receptor function, cellular metabolism, circadian cycle and epigenetics. KIAA1967 acts as a regulator of various nuclear receptors such as ERα (oestrogen receptor α), ERβ, AR (androgen receptor), RAR (retinoic acid receptor) and haem receptor Rev-erbα. In addition to transcriptional activity, it also controls epigenetic modifiers such as the deacetylases HDAC3 and SIRT1, and the methyl-transferase SUV39H1. It has been reported that Nudix domain has the ability to bind nucleotides, which further could control its functional activities. In response to UV-induced DNA damage, it plays an important role in the maintenance of genomic stability and cellular integrity. KIAA1967 also plays a significant role in adipocyte differentiation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74217

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José María Moreno-Navarrete et al.
American journal of physiology. Endocrinology and metabolism, 308(7), E554-E561 (2015-02-05)
Genetic deletion of Dbc1 in mice reduced adipose tissue senescence and inflammation while promoting an expansion of this tissue. Here, we aimed to investigate DBC1 mRNA and protein levels in human adipose tissue from subjects with a wide spectrum of
Eduardo Nunes Chini et al.
Bioscience reports, 33(4), doi:10-doi:10 (2013-07-12)
DBC1 (deleted in breast cancer-1) is a nuclear protein that regulates cellular metabolism. Since alteration in cellular metabolism have been proposed to be the emerging 'hallmark' of cancer, it is possible that DBC1 may be implicated in the regulation of
Wootae Kim et al.
Cancer letters, 333(2), 180-186 (2013-01-29)
DBC1 (deleted in breast cancer 1) participates in the regulation of cell survival and death in response to various stimuli. In particular, DBC1 promotes cell death upon DNA damage through inhibition of SIRT1 deacetylase. However, the SIRT1-independent functions of DBC1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service