Skip to Content
Merck
All Photos(8)

Key Documents

HPA019827

Sigma-Aldrich

Anti-NUDT5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ADP-sugar pyrophosphatase, Anti-Nucleoside diphosphate-linked moiety X motif 5, Anti-Nudix motif 5, Anti-YSA1H

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SPAVCMDPGLSNCTIHIVTVTINGDDAENARPKPKPGDGEFVEVISLPKNDLLQRLDALVAEEHLTVDARVYSYALALKHANAKPFEVPFLKF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NUDT5(11164)

General description

NUDT5 (Nudix, nucleoside diphosphate linked moiety X-type motif 5) is an ADP-ribose pyrophosphatase (ADPRase) consisting of an N-terminal domain with three-stranded antiparallel β-sheet named β1, β2, and β3, the catalytic Nudix domain, and an extended C-terminal α-helix domain. It is a novel member of the Nudix hydrolase superfamily and exists as a homodimer.

Immunogen

ADP-sugar pyrophosphatase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NUDT5 (Nudix, nucleoside diphosphate linked moiety X-type motif 5) is mainly involved in maintaining the intracellular level of ADP-ribose by hydrolyzing ADP-ribose (ADPR) to AMP and ribose 5′-phosphate. It also restricts non-enzymatic ADP-ribosylation. Its N-terminal antiparallel β-sheet plays an important role in dimerization, which is essential for substrate recognition and enzymatic activity. NUDT5 also has been predicted for altered cell proliferation. It has been suggested that suppressed NUDT5 cell shows delayed cell cycle G1 phase. Thus, it has been concluded that NUDT5 may be involved in cell growth and cell cycle proliferation by regulating G1-S transition in mammalian cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74713

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manwu Zha et al.
Journal of molecular biology, 364(5), 1021-1033 (2006-10-21)
Human NUDT5 (hNUDT5) is an ADP-ribose pyrophosphatase (ADPRase) belonging to the Nudix hydrolase superfamily. It presumably plays important roles in controlling the intracellular level of ADP-ribose (ADPR) to prevent non-enzymatic ADP-ribosylation by hydrolyzing ADPR to AMP and ribose 5'-phosphate. We
Li-Qun Zhang et al.
Molecular and cellular biochemistry, 363(1-2), 377-384 (2011-12-28)
The molecule 8-oxo-7,8-dihydroguanine (8-oxoGua), an oxidized form of guanine, can pair with adenine or cytosine during nucleic acid synthesis. Moreover, RNA containing 8-oxoGua causes translational errors, thus leading to the production of abnormal proteins. Human NUDT5, a MutT-related protein, catalyzes
Hong-Nu Yu et al.
Biochemical and biophysical research communications, 354(3), 764-768 (2007-01-31)
The ADP-ribose (ADPR) pyrophosphatase (ADPRase) NUDT5, a member of a superfamily of Nudix hydrolases, hydrolyzes ADP-ribose (ADPR) to AMP and ribose 5'-phosphate. Nitric oxide (NO) enhances nonenzymatic ADP-ribosylation of proteins such as beta-actin and glyceraldehydes 3-phosphate dehydrogenase in the presence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service