Skip to Content
Merck
All Photos(8)

Key Documents

HPA019713

Sigma-Aldrich

Anti-NCS1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FREQ, Anti-Frequenin homolog, Anti-Frequenin-like protein, Anti-Frequenin-like ubiquitous protein, Anti-NCS-1, Anti-Neuronal calcium sensor 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

YITRNEMLDIVDAIYQMVGNTVELPEEENTPEKRVDRIFAMMDKNADGKLTLQEFQEGSKADPSIVQALSLYDGLV

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FREQ(23413)

General description

NCS1 (Neuronal calcium sensor 1) is a multispecific, calcium sensor belonging to the EF-hand calcium-binding protein superfamily. It is expressed in postsynaptic sites, such as somata and dendrites of Purkinje cells, stellate/basket cells and granule cells.

Immunogen

Neuronal calcium sensor 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NCS1 (Neuronal calcium sensor 1) is involved in series of neuronal activities such as controlling of synaptic transmission and plasticity. It also plays a crucial role in the release of neurotransmitter molecules, which has clinical correlation with neurological diseases. It also has been categorized as dopamine receptor-interacting proteins (DRIPs) since it has relation to the susceptibility and pathophysiology of cocaine addiction (CA).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75196

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shozo Jinno et al.
Cerebellum (London, England), 3(2), 83-88 (2004-07-06)
Neuronal calcium sensor-1 (NCS-1) is a member of EF-hand calcium-binding protein superfamily, which is considered to modulate synaptic transmission and plasticity. In this mini-review, we first summarize distribution of NCS-1 in the cerebellum. NCS-1 is mainly detected in postsynaptic sites
Pétur O Heidarsson et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(36), 13069-13074 (2014-08-27)
Neurodegenerative disorders are strongly linked to protein misfolding, and crucial to their explication is a detailed understanding of the underlying structural rearrangements and pathways that govern the formation of misfolded states. Here we use single-molecule optical tweezers to monitor misfolding
S Averill et al.
Neuroscience, 123(2), 419-427 (2003-12-31)
Neuronal calcium sensor-1 (NCS-1) is a member of the EF-hand calcium-binding protein superfamily which has been implicated in the modulation of a number of neuronal functions. In this study we have examined the expression of NCS-1 in adult rat dorsal
Pushpinder K Multani et al.
Neuroscience letters, 531(1), 46-51 (2012-09-25)
Genes involved in drug reward pathways are plausible candidates for susceptibility to substance use disorders. Given the prominent role of dopamine in drug reward, dopamine receptor-interacting proteins (DRIPs) such as the neuronal calcium sensor-1 (NCS-1) protein have been hypothesized to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service