Skip to Content
Merck
All Photos(8)

Key Documents

HPA019678

Sigma-Aldrich

Anti-UCHL3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-UCH-L3, Anti-Ubiquitin carboxyl-terminal hydrolase isozyme L3, Anti-Ubiquitin thioesterase L3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

MEGQRWLPLEANPEVTNQFLKQLGLHPNWQFVDVYGMDPELLSMVPRPVCAVLLLFPITEKYEVFRTEEEEKIKSQGQDVTSSVYFMKQTISN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UCHL3(7347)

General description

Ubiquitin carboxyl-terminal esterase L3 (UCH-L3) belongs to the category of deubiquitinating enzymes (DUBs),and is expressed in the brain. The tissue-specific protein has a molecular mass of 26.182Da.

Immunogen

Ubiquitin carboxyl-terminal hydrolase isozyme L3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Ubiquitin carboxyl-terminal esterase L3 (UCH-L3) is mainly involved in the regulation of central nervous system. It separates ubiquitin (Ub) from Ub precursors or protein substrates. UCH-L3 hydrolyzes C-terminal extension of mutant ubiquitin.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74693

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frank J A Dennissen et al.
FEBS letters, 585(16), 2568-2574 (2011-07-19)
Mutant ubiquitin (UBB(+1)) accumulates in the hallmarks of tauopathies and polyglutamine diseases. We show that the deubiquitinating enzyme YUH1 of Saccharomyces cerevisiae and its mouse and human ortholog UCH-L3 are able to hydrolyze the C-terminal extension of UBB(+1). This yields
Hyun Min Song et al.
Biochemical and biophysical research communications, 452(3), 722-727 (2014-09-10)
Ubiquitin C-terminal hydrolase-L3 (UCH-L3) is among the deubiquitinating enzymes (DUBs) that cleave ubiquitin (Ub) from Ub precursors or protein substrates. Many DUBs have been shown to participate in cancer progression in various tissues. However, the mechanism and role of UCH-L3
Rieko Setsuie et al.
Neurochemistry international, 54(5-6), 314-321 (2009-01-22)
Ubiquitin (Ub) carboxy terminal hydrolase (UCH)-L1 and UCH-L3 are two of the deubiquitinating enzymes expressed in the brain. Both gad mice, which lack UCH-L1 expression and Uchl3 knockout mice exhibit neurodegeneration, although at distinct areas. These phenotypes indicate the importance
K D Wilkinson et al.
Science (New York, N.Y.), 246(4930), 670-673 (1989-11-03)
A complementary DNA (cDNA) for ubiquitin carboxyl-terminal hydrolase isozyme L3 was cloned from human B cells. The cDNA encodes a protein of 230 amino acids with a molecular mass of 26.182 daltons. The human protein is very similar to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service