Skip to Content
Merck
All Photos(5)

Key Documents

HPA019445

Sigma-Aldrich

Anti-KCNH1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Ether-a-go-go potassium channel 1, Anti-Potassium voltage-gated channel subfamily H member 1, Anti-Voltage-gated potassium channel subunit Kv10.1, Anti-h-eag, Anti-hEAG1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

DSCDSGITKSDLRLDNVGEARSPQDRSPILAEVKHSFYPIPEQTLQATVLEVRHELKEDIKALNAKMTNIEKQLSEILRILTSRRSSQSPQELFEISRPQSPES

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNH1(3756)

General description

KCNH1 (potassium voltage-gated channel subfamily H member 1) is mainly expressed in the central nervous system, but is also present in myoblasts, adrenal gland, placenta and testis. The protein localizes at the cell surface, early endosomes and the inner nuclear membrane. KCNH1 contains transmembrane domains, calmodulin binding site, PAS (Per-Arnt-Sim) domain, cyclic nucleotide binding domain and a nuclear localization signal. KCNH1 is also referred to as EAG1 (ether a go-go1).

Immunogen

Potassium voltage-gated channel subfamily H member 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

KCNH1 (potassium voltage-gated channel subfamily H member 1) is a voltage-gated potassium channel. Mutations in KCNH1 are associated with Zimmermann-Laband syndrome, Temple-Baraitser syndrome and epilepsy. KCNH1 is up-regulated in various cancers. It regulates cell cycle and proliferation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73126

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Bronk et al.
Journal of neurophysiology, 119(5), 1665-1680 (2018-01-25)
Drosophila ether-à-go-go ( eag) is the founding member of a large family of voltage-gated K+ channels, the KCNH family, which includes Kv10, 11, and 12. Concurrent binding of calcium/calmodulin (Ca2+/CaM) to NH2- and COOH-terminal sites inhibits mammalian EAG1 channels at
Cas Simons et al.
Nature genetics, 47(1), 73-77 (2014-11-25)
Temple-Baraitser syndrome (TBS) is a multisystem developmental disorder characterized by intellectual disability, epilepsy, and hypoplasia or aplasia of the nails of the thumb and great toe. Here we report damaging de novo mutations in KCNH1 (encoding a protein called ether
Jin Wu et al.
BioMed research international, 2014, 345678-345678 (2014-08-20)
The ether à go-go1 (Eag1) channel is overexpressed in a variety of cancers. However, the expression and function of Eag1 in liposarcoma are poorly understood. In the present study, the mRNA expression of Eag1 in different adipose tissue samples was
Fanny Kortüm et al.
Nature genetics, 47(6), 661-667 (2015-04-29)
Zimmermann-Laband syndrome (ZLS) is a developmental disorder characterized by facial dysmorphism with gingival enlargement, intellectual disability, hypoplasia or aplasia of nails and terminal phalanges, and hypertrichosis. We report that heterozygous missense mutations in KCNH1 account for a considerable proportion of
Janice García-Quiroz et al.
Revista de investigacion clinica; organo del Hospital de Enfermedades de la Nutricion, 71(3), 186-194 (2019-06-12)
Expression and activity of the potassium channel ether-à-go-go-1 (EAG1) are strongly related to carcinogenesis and tumor progression, which can be exploited for therapeutic purposes. EAG1 activity may be reduced by preventing its phosphorylation with epidermal growth factor receptor (EGFR) kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service