Skip to Content
Merck
All Photos(2)

Key Documents

HPA019367

Sigma-Aldrich

Anti-KRT75 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CK-75, Anti-Cytokeratin-75, Anti-K75, Anti-Keratin, type II cytoskeletal 75, Anti-Keratin-75, Anti-Type II keratin-18, Anti-hK6hf

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

MSRQSSITFQSGSRRGFSTTSAITPAAGRSRFSSVSVARSAAGSGGLGRISSA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT75(9119)

General description

The gene KRT75 (keratin, type II cytoskeletal 75) is mapped to human chromosome 12q13. The protein localizes in the companion layer of the hair follicle, upper matrix and medulla of the anagen-stage hair. The expression of KRT75 is controlled in a hair-cycle-dependent manner. It is absent in the telogen phase of the hair cycle. KRT75 is also referred to as K6HF (keratin-6 hair follicle).

Immunogen

Keratin, type II cytoskeletal 75 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Mutations in KRT75 (keratin, type II cytoskeletal 75) are linked with loose anagen hair syndrome. It is up-regulated in dermal papilla-induced keratinocyte differentiation. In mice, polymorphism in KRT75 which causes hair disorders also results in increased dental caries.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74767

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiling Wang et al.
The Journal of investigative dermatology, 121(6), 1276-1282 (2003-12-17)
More than half of the known keratin genes (n approximately 50) are expressed in the hair follicle. An in-depth knowledge of their differential expression in this organ will help us to understand the mechanisms of its formation and cycling, and
Olivier Duverger et al.
The Journal of clinical investigation, 124(12), 5219-5224 (2014-10-28)
Tooth enamel is the hardest substance in the human body and has a unique combination of hardness and fracture toughness that protects teeth from dental caries, the most common chronic disease worldwide. In addition to a high mineral content, tooth
Ichiro Kurokawa et al.
Experimental dermatology, 20(3), 217-228 (2011-02-18)
Keratins are a highly diverse family of cytoskeletal proteins and important markers of epithelial cell differentiation. In this review, applying the new keratin nomenclature recently introduced, we summarize and discuss the distribution and significance of keratin patterns in cutaneous epithelial
H Winter et al.
The Journal of investigative dermatology, 111(6), 955-962 (1998-12-18)
In an attempt to identify new members of the human type II hair keratin family by means of 3'- and 5'-RACE methods and cDNA from anagen hair follicles, we detected a sequence that encoded a hitherto unknown type II cytokeratin.
Valérie Chapalain et al.
Archives of dermatology, 138(4), 501-506 (2002-04-10)
To report the clinical features of the loose anagen hair syndrome and to test the hypothesis that the typical gap between the hair and the inner root sheath may result from hereditary defects in the inner root sheath or the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service