Skip to Content
Merck
All Photos(2)

Key Documents

HPA019263

Sigma-Aldrich

Anti-PPM1E antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-CaMKN, Anti-CaMKP-N, Anti-CaMKP-nucleus, Anti-Partner of PIX 1, Anti-Partner of PIXA, Anti-Protein phosphatase 1E

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

TVIVVFLRDMNKAVNVSEESDWTENSFQGGQEDGGDDKENHGECKRPWPQHQCSAPADLGYDGRVDSFTDRTSLSPGSQINVLE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPM1E(22843)

General description

The gene PPM1E (protein phosphatase Mg2+/Mn2+ dependent 1E) is mapped to human chromosome 17q22. It belongs to PPM family. PPM1E is abundantly expressed in brain and testis.

Immunogen

Protein phosphatase 1E recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The autophosphorylation of p21 (CDC42/Rac)-activated kinase PAK is crucial for mediating cellular functions. The protein phosphatase Mg2+/Mn2+ dependent 1E (PPM1E) interacts with the PAK (p21-activated kinase) interacting guanine nucleotide exchange factor (PIX). The dephosphrylation action of PPM1E suppresses active PAK responses. PPM1E is also a phosphatase for 5′-AMP-activated protein kinase (AMPK). Activation and phosphorylation of AMPK is important for treatment of type-2 diabetes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73653

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Voss et al.
Cellular signalling, 23(1), 114-124 (2010-08-31)
Activation of 5'-AMP-activated protein kinase (AMPK) is believed to be the mechanism by which the pharmaceuticals, metformin and phenformin, exert their beneficial effects for treatment of type 2 diabetes. These biguanide drugs elevate 5'-AMP, which allosterically activates AMPK and promotes
K Litchfield et al.
Andrology, 3(1), 34-46 (2015-01-20)
Testicular germ cell tumour (TGCT) is the most common cause of cancer in young men (aged 15-45 years) in many populations. Multiple genome-wide association studies (GWAS) of TGCT have now been conducted, yielding over 25 disease-associated single-nucleotide polymorphism (SNP)s at 19
Wenjing Sun et al.
The Journal of biological chemistry, 285(28), 21341-21348 (2010-05-08)
MEKK3 is a central intermediate signaling component in lysophosphatidic acid (LPA)-induced activation of the nuclear factor-kappaB (NF-kappaB). However, the precise mechanism for the termination of MEKK3 kinase activity is not fully understood. Using a functional genomic approach, we have identified
Cheng-Gee Koh et al.
Current biology : CB, 12(4), 317-321 (2002-02-28)
The Rho GTPases are involved in many signaling pathways and cellular functions, including the organization of the actin cytoskeleton, regulation of transcription, cell motility, and cell division. The p21 (Cdc42/Rac)-activated kinase PAK mediates a number of biological effects downstream of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service