Skip to Content
Merck
All Photos(8)

Key Documents

HPA019206

Sigma-Aldrich

Anti-AQP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AQP-1, Anti-Aquaporin-1, Anti-Aquaporin-CHIP, Anti-Urine water channel, Anti-Water channel protein for red blood cells and kidney proximal tubule

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Check Cart for Availability


Select a Size

Change View
100 μL
€505.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

PRSSDLTDRVKVWTSGQVEEYDLDADDINSRVEMKPK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AQP1(358)

General description

The gene AQP1 (aquaporin-1) is mapped to human chromosome 7p14. It is an integral membrane protein.

Immunogen

Aquaporin-1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

AQP1 (aquaporin-1) is responsible for osmotic water permeability. Co-expression of AQP1 with carbonic anhydrase II (CAII) increases plasma membrane water permeability.[1] Down-regulation of AQP1 is associated with lumbar intervertebral disc degeneration. AQP1 is also linked with portal hypertension in primary biliary cirrhosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73099

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gonzalo Vilas et al.
Molecular biology of the cell, 26(6), 1106-1118 (2015-01-23)
Aquaporin-1 (AQP1) enables greatly enhanced water flux across plasma membranes. The cytosolic carboxy terminus of AQP1 has two acidic motifs homologous to known carbonic anhydrase II (CAII) binding sequences. CAII colocalizes with AQP1 in the renal proximal tubule. Expression of
Shirin Shahbazi et al.
Medicine, 94(48), e2144-e2144 (2015-12-04)
Hematological parameters are appraised routinely to determine overall human health and to diagnose and monitor certain diseases. In GWASs, more than 30 loci carrying common deoxyribonucleic acid (DNA) polymorphisms have been identified related to hematological traits. In this study, we
Hiroyoshi Iguchi et al.
Medical molecular morphology, 47(2), 90-99 (2013-08-21)
Although aquaporins (AQPs) in normal hepatobiliary system have been studied, little is known about AQP localization and changes in the hepatic microvascular system including sinusoids in cholestatic liver. The present study aimed to clarify the localization of AQP-1 in the
S B Li et al.
Genetics and molecular research : GMR, 13(4), 8225-8233 (2014-10-10)
Lumbar intervertebral disc degeneration is induced by multiple factors, but few studies have examined the effects of aquaporins on this process. We compared the expression levels of aquaporins 1 and 3 in normal and degenerative lumbar intervertebral discs. Fifteen normal
G M Preston et al.
Science (New York, N.Y.), 256(5055), 385-387 (1992-04-17)
Water rapidly crosses the plasma membrane of red blood cells (RBCs) and renal tubules through specialized channels. Although selective for water, the molecular structure of these channels is unknown. The CHIP28 protein is an abundant integral membrane protein in mammalian

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service