Skip to Content
Merck
All Photos(8)

Documents

HPA019182

Sigma-Aldrich

Anti-TBL1XR1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-F-box-like/WD repeat-containing protein TBL1XR1, Anti-Nuclear receptor corepressor/HDAC3 complex subunit TBLR1, Anti-TBL1-related protein 1, Anti-Transducin beta-like 1X-related protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

RPIESLSLIDAVMPDVVQTRQQAYRDKLAQQQAAAAAAAAAAASQQGSAKNGENTANGEENGAHTIANNHTDMMEVDGDVEIPPNKAVVLRGHE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TBL1XR1(79718)

General description

The gene TBL1XR1 (transducin β-like 1X-related protein 1) is mapped to human chromosome 3q26.32. The protein contains four WD40 (β-transducin) repeats. TBL1XR1 is also referred as TBLR1 (TBL1-related protein 1).

Immunogen

F-box-like/WD repeat-containing protein TBL1XR1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TBL1XR1 (transducin β-like 1X-related protein 1) regulates the change from gene repression to activation of nuclear receptors and proteins involved in transcriptional activation. It also functions as an E3-ligase. TBL1XR1 haploinsufficiency results in intellectual disability, without autistic behavior. Mutations in TBL1XR1 are linked with autism spectrum disorders. TBL1XR1 is involved in progression of breast cancer cells by transactivation of cyclin D1 and activation of the β-catenin signaling pathway.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72965

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinghua Li et al.
Breast cancer research : BCR, 16(5), 465-465 (2014-10-25)
Transducin (β)-like 1 X-linked receptor 1(TBLR1) is an F-box-like and WD repeat-containing protein which functions as a switch in transcriptional activation, However, the clinical significance and biological role of TBLR1 in breast cancer remains largely unknown. Western blotting, immunocytochemistry and
Hongda Liu et al.
Scientific reports, 7, 44275-44275 (2017-03-16)
More than 25% localized CRC patients died from post-operative metastasis, and risk of metastasis varies among individuals due to the high heterogeneity of CRC. Therefore, figuring out potential biomarkers for disease recurrence would be invaluable to improve the follow-up efficiency
Linda Pons et al.
American journal of medical genetics. Part A, 167A(1), 164-168 (2014-11-27)
We report here on an 8-year-old girl and her mother, both displaying similar facial dysmorphism, speech delay, and mild to moderate intellectual disability. Array-CGH studies revealed the same interstitial 3q26.32 microdeletion encompassing a single coding gene, TBL1XR1, in both patients.
Hongda Liu et al.
Journal of gastroenterology and hepatology, 32(9), 1570-1580 (2017-01-28)
A considerable number of early-stage colorectal cancer (CRC) patients may develop cancer relapse or metastasis after curative surgery. Isolated tumor cells (ITC) and micrometastasis in lymph nodes (LNMM), which are undetectable by conventional pathological examination, may be one primary reason.

Articles

Learn about glioma markers for high-grade gliomas and low-grade gliomas and find reliable Prestige Antibodies® to target glioma markers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service