Skip to Content
Merck
All Photos(5)

Key Documents

HPA019150

Sigma-Aldrich

Anti-CREB1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CREB, Anti-cAMP response element-binding protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€482.00

€482.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€482.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€482.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human, mouse

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

STIAESEDSQESVDSVTDSQKRREILSRRPSYRKILNDLSSDAPGVPRIEEEKSEEETSAPAITTVTVPTPIYQTSSGQYIAITQGGAIQLANNGTDGVQG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CREB1(1385)

General description

The gene CREB1 (cyclic AMP-responsive element-binding protein 1) is mapped to human chromosome 2q34. It is a member of the leucine zipper family of DNA-binding proteins. CREB1 is widely expressed.[1]

Immunogen

cAMP response element-binding protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CREB1 (cyclic AMP-responsive element-binding protein 1) is a cAMP (cyclic adenosine monophosphate) responsive transcription factor which is important for neuronal signal transduction. It is involved in cell division and apoptosis.[2] CREB1 is phosphorylated by cAMP-dependent protein kinase A, Ca2+-calmodulin, AKT (RAC-α serine/threonine-protein kinase), p38 MAPK (mitogen-activated protein kinase 1), JNK (c-Jun N-terminal kinase), and other kinases. Phosphorylation increases the transcriptional activity of CREB1 and its association with the co-activator, CREB-binding protein (CBP).[1][2] Polymorphism in CREB1 is associated with major depressive disorder. It also participates in Kaposi′s Sarcoma-associated herpesvirus lytic replication during primary infection. CREB1 is also linked with schizophrenia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86544

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Ma et al.
Journal of psychiatric research, 57, 84-89 (2014-07-22)
CREB1 is a cAMP responsive transcriptional factor which plays a key role in neural development. CREB1 signal pathway (CSP) has been implicated repeatedly in studies of predisposition for schizophrenia. We speculated that CSP has undergone positive selection during evolution of
Peng Wang et al.
International journal of clinical and experimental pathology, 8(1), 906-913 (2015-03-11)
Major depressive disorder (MDD) is one of the most severe psychiatric disorders. The objective of this study was to explore the effects of CREB1 gene polymorphisms on risk of developing MDD and the joint effects of gene-environment interactions. Genotyping was
Shun-Fa Yang et al.
Oncotarget, 6(5), 2736-2753 (2015-01-22)
Osteosarcomas, the most common malignant bone tumors, show a potent capacity for local invasion and pulmonary metastasis. Resveratrol (RESV), a phytochemical, exhibits multiple tumor-suppressing activities and has been tested in clinical trials. However, the antitumor activities of RESV in osteosarcomas
H-D Chae et al.
Leukemia, 29(6), 1379-1389 (2014-12-30)
CREB (cyclic AMP response element-binding protein) is a transcription factor overexpressed in normal and neoplastic myelopoiesis and regulates cell cycle progression, although its oncogenic mechanism has not been well characterized. Replication factor C3 (RFC3) is required for chromatin loading of
Fan Cheng et al.
Journal of virology, 89(18), 9262-9280 (2015-06-26)
Viruses often hijack cellular pathways to facilitate infection and replication. Kaposi's sarcoma-associated herpesvirus (KSHV) is an oncogenic gammaherpesvirus etiologically associated with Kaposi's sarcoma, a vascular tumor of endothelial cells. Despite intensive studies, cellular pathways mediating KSHV infection and replication are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service