Skip to Content
Merck
All Photos(2)

Key Documents

HPA019018

Sigma-Aldrich

Anti-MCTP1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Multiple C2 and transmembrane domain-containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

VFDYDFGLQDDFMGSAFLDLTQLELNRPTDVTLTLKDPHYPDHDLGIILLSVILTPKEGESRDVTMLMRKSWKRSSKFQTQSLRLSDLHRKSHLWRGIVSITLIEGRDLKAMDSNGLSDPYVKFRLGH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MCTP1(79772)

General description

The gene MCTP1 (multiple C2 and transmembrane domain-containing protein 1) is mapped to human chromosome 5q15. It belongs to MCTP family of conserved C2 domain proteins. MCTP1 is strongly expressed in skeletal and heart muscle. The protein localizes at the cell membrane.

Immunogen

Multiple C2 and transmembrane domain-containing protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MCTP1 (Multiple C2 and transmembrane domain-containing protein 1) participates in Ca2+-associated signaling process. It is suggested to be linked with Alzheimer′s disease. Similarly, single nucleotide polymorphism in MCTP1 is linked with bipolar disorder. MCTP1 is also proposed to regulate endocytic recycling of specific neurons and synapses.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72448

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lifeng Qiu et al.
Journal of neurochemistry, 135(3), 492-507 (2015-07-22)
Multiple C2 domains transmembrane protein 1 (MCTP1) contains two transmembrane regions and three C2 domains of high Ca(2+)-binding affinity. Single-nucleotide polymorphism (SNP) of human MCTP1 gene is reportedly associated with bipolar disorder, but expression and function of MCTP1 in the
Margherita Squillario et al.
BMC medical genomics, 4, 55-55 (2011-07-06)
A molecular characterization of Alzheimer's Disease (AD) is the key to the identification of altered gene sets that lead to AD progression. We rely on the assumption that candidate marker genes for a given disease belong to specific pathogenic pathways
Ok-Ho Shin et al.
The Journal of biological chemistry, 280(2), 1641-1651 (2004-11-06)
C2 domains are primarily found in signal transduction proteins such as protein kinase C, which generally contain a single C2 domain, and in membrane trafficking proteins such as synaptotagmins, which generally contain multiple C2 domains. In both classes of proteins
Laura J Scott et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(18), 7501-7506 (2009-05-07)
Bipolar disorder (BP) is a disabling and often life-threatening disorder that affects approximately 1% of the population worldwide. To identify genetic variants that increase the risk of BP, we genotyped on the Illumina HumanHap550 Beadchip 2,076 bipolar cases and 1,676

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service