Skip to Content
Merck
All Photos(6)

Documents

HPA018840

Sigma-Aldrich

Anti-FHIT antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AP3A hydrolase, Anti-AP3Aase, Anti-Bis(5′-adenosyl)-triphosphatase, Anti-Diadenosine 5′,5″′- P1,P3-triphosphate hydrolase, Anti-Dinucleosidetriphosphatase, Anti-Fragile histidine triad protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

FRFGQHLIKPSVVFLKTELSFALVNRKPVVPGHVLVCPLRPVERFHDLRPDEVADLFQTTQRVGTVVE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FHIT(2272)

General description

The gene FHIT (fragile histidine triad protein) is mapped to human chromosome 3p14.2. It belongs to histidine-triad superfamily of nucleoside monophosphate hydrolases and transferases. The protein is present in the cytoplasm and the nucleus.

Immunogen

Bis(5′-adenosyl)-triphosphatase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Fragile histidine triad protein (FHIT) is a dinucleoside 5′,5′′′-P1,P3-triphosphate hydrolase. It is a tumor suppressor gene. It is down-regulated in cervical cancer and non-small cell lung cancer. Co-expression of FHIT and p53 inhibits cell proliferation in non-small cell lung carcinoma. Absence of FHIT induces the expression of oxidative stress response genes after contact with cigarette smoke extract.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74606

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco Trapasso et al.
Proceedings of the National Academy of Sciences of the United States of America, 100(4), 1592-1597 (2003-02-08)
The FHIT gene is inactivated early in the development of many human tumors, and Fhit-deficient mice have increased cancer incidence. Viral reexpression of Fhit kills Fhit-deficient cells by induction of apoptosis. Fhit, a member of branch 2 of the histidine-triad
Masahiko Nishizaki et al.
Cancer research, 64(16), 5745-5752 (2004-08-18)
Aberrations of the tumor suppressor genes FHIT and p53 are frequently associated with a wide range of human cancers, including lung cancer. We studied the combined effects of FHIT and p53 proteins on tumor cell proliferation and apoptosis in human
Li-Xia Bai et al.
Asian Pacific journal of cancer prevention : APJCP, 15(21), 9313-9317 (2014-11-26)
Fragile histidine triad (FHIT) is a suppressor gene related to cervical cancer through CpG island hypermethylation. Folate is a water-soluble B-vitamin and an important cofactor in one-carbon metabolism. It may play an essential role in cervical lesions through effects on
Roberto Jose Diaz et al.
Neoplasia (New York, N.Y.), 14(9), 788-798 (2012-09-29)
Chordoma is a rare tumor arising in the sacrum, clivus, or vertebrae. It is often not completely resectable and shows a high incidence of recurrence and progression with shortened patient survival and impaired quality of life. Chemotherapeutic options are limited
D-W Wu et al.
Oncogene, 34(16), 2072-2082 (2014-06-10)
The dual role of the microRNA-29 (miR-29) family in tumor progression and metastasis in solid tumors has been reported. Evidence for the role of miR-29 in tumor malignancy and its prognostic value in overall survival (OS) and relapse-free survival (RFS)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service