Skip to Content
Merck
All Photos(3)

Key Documents

HPA018524

Sigma-Aldrich

Anti-NDUFS8 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CI-23kD, Anti-Complex I-23kD, Anti-NADH dehydrogenase [ubiquinone] iron-sulfur protein 8, mitochondrial precursor, Anti-NADH-ubiquinone oxidoreductase 23 kDa subunit, Anti-TYKY subunit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

AATYKYVNMQDPEMDMKSVTDRAARTLLWTELFRGLGMTLSYLFREPATINYPFEKGPLSPRFRGEHALRRYPSGEERCIACKLCEAICPAQAITIEAEPRADGSRRTTRYDIDMTKCIYC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NDUFS8(4728)

General description

The gene NDUFS8 (NADH dehydrogenase [ubiquinone] iron-sulfur protein 8 - mitochondrial) is mapped to human chromosome 11q13. It is a nuclear encoded gene and is localized in the mitochondria. The protein contains two 4Fe-4S ferredoxin consensus patterns.

Immunogen

NADH dehydrogenase [ubiquinone] iron-sulfur protein 8, mitochondrial precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NADH dehydrogenase [ubiquinone] iron-sulfur protein 8 - mitochondrial (NDUFS8) is a subunit of nicotinamide adenine dinucleotide (NADH):ubiquinone oxidoreductase (complex I) enzyme complex of the respiratory chain. Mutations in NDUFS8 are associated with Leigh syndrome. NDUFS8 is down-regulated in adrenocortical carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86627

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gustavo G Fernandez-Ranvier et al.
World journal of surgery, 32(5), 873-881 (2008-03-08)
The most common genetic change observed in adrenocortical carcinoma is loss of heterozygozity on chromosome 11q13. As genes on this chromosome may be important in the pathogenesis of adrenocortical carcinoma, we compared their expression profile between benign and malignant adrenocortical
J Loeffen et al.
American journal of human genetics, 63(6), 1598-1608 (1998-12-05)
Nicotinamide adenine dinucleotide (NADH):ubiquinone oxidoreductase (complex I) is the largest multiprotein enzyme complex of the respiratory chain. The nuclear-encoded NDUFS8 (TYKY) subunit of complex I is highly conserved among eukaryotes and prokaryotes and contains two 4Fe4S ferredoxin consensus patterns, which
Vincent Procaccio et al.
Neurology, 62(10), 1899-1901 (2004-05-26)
Analysis of the complex I NDUFS8 gene from Leigh syndrome patients with isolated complex I deficiency revealed that one patient with late-onset disease and partial complex I defect was a compound heterozygote for two novel mutations in NDUFS8 gene. Western

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service