Skip to Content
Merck
All Photos(3)

Key Documents

HPA018460

Sigma-Aldrich

Anti-DSCR4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Down syndrome critical region protein 4, Anti-Down syndrome critical region protein B

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

MSLIILTRDDEPRIFTPDSDAASPALHSTSPLPDPASASPLHREEKILPKVCNIVSCLSFSLPASPTDSGLASPTIITREGQQFWAKCLIWKYQLYLHGLHKKSDGR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DSCR4(10281)

General description

The gene DSCR4 (Down syndrome critical region protein-4) is mapped to human chromosome 21q22.2. DSCR4 is restricted to human placenta.

Immunogen

Down syndrome critical region protein 4 recombinant protein epitope signature tag (PrEST)

Application

Anti-DSCR4 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Down syndrome critical region protein-4 (DSCR4) promoter is shown to be a fetal epigenetic marker for noninvasive prenatal detection of chromosomal aberration trisomy 21. DSCR4 promoter is hypomethylated in placental and densely methylated in maternal blood cells. However, unmethylated DSCR4 can be identified in the first-trimester maternal plasma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73904

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoko Asai et al.
Biochimica et biophysica acta, 1779(1), 40-50 (2007-12-19)
The gene DSCR4 locates in the band q22.2 of human chromosome 21 and encodes a protein of 118 amino acids. Expression of DSCR4 is restricted to human placenta and placental choriocarcinoma cell lines BeWo and JEG3. The 5'-RACE method using
Yingying Du et al.
Prenatal diagnosis, 31(2), 207-214 (2011-01-27)
Trisomy 21 is the most common chromosomal aberration in live births. Some efforts have been made to develop noninvasive prenatal detection of trisomy 21 by using fetal DNA in maternal plasma. Due to the maternal DNA background, a distinguishable marker
Atsushi Toyoda et al.
Genome research, 12(9), 1323-1332 (2002-09-06)
Comprehensive knowledge of the gene content of human chromosome 21 (HSA21) is essential for understanding the etiology of Down syndrome (DS). Here we report the largest comparison of finished mouse and human sequence to date for a 1.35-Mb region of
Mathias Uhlén et al.
Molecular & cellular proteomics : MCP, 11(3), M111-M111 (2011-11-02)
The Human Proteome Project has been proposed to create a knowledge-based resource based on a systematical mapping of all human proteins, chromosome by chromosome, in a gene-centric manner. With this background, we here describe the systematic analysis of chromosome 21

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service