Skip to Content
Merck
All Photos(7)

Key Documents

HPA018156

Sigma-Aldrich

Anti-CTSB antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-APP secretase, Anti-APPS, Anti-Cathepsin B precursor, Anti-Cathepsin B1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

DELVNYVNKRNTTWQAGHNFYNVDMSYLKRLCGTFLGGPKPPQRVMFTEDLKLPASFDAREQWPQCPTIKEIRDQGSCGSCWAFGAVEAISDRICIHTNAHVSVEVSAEDLLTCCGSMCGDGCNGGYPA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTSB(1508)

General description

The gene cathepsin B (CTSB) has been mapped to human chromosome 8p22. The gene encodes a lysosomal cysteine protease composed of a dimer of disulphide linked heavy and light chains. Under physiological conditions the protein is localized in lysosomes and in presence of some cellular signals it is released from the lysosomes and trigger different physiological events.[1]

Immunogen

Cathepsin B precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-CTSB antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Cathepsin B (CTSB) has been linked to various responses including lysosomal protein turnover, apoptosis and immune responses. The protein is involved in activation of epithelial Na+ channel in cystic fibrosis lung disease airways. Presence of cocaine along with HIV (human immunodeficiency virus)-1 infection causes increase in CTSB secretion and neurotoxicity. CTSB is up-regulated in synovial fluid of patients suffering with rheumatoid arthritis. The protein is considered as a biomarker for many cancers. Stearoyl-CoA desaturase-5 reduces melanoma malignancy by regulating secretion of CTSB. Mutations in CTSB (C76G/A4383C) have been linked to high risk of hepatocellular carcinoma. CTSB levels and activity were shown to be increased in human esophageal squamous cell carcinoma, breast cancer, colorectal carcinomas and no small cell lung carcinoma.[1][2] CTSB increase the localization of cytoplasmic c-Jun-NH2-terminus kinase, thereby inducing migration of glioma cells and making them more malignant.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73333

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Ma et al.
PloS one, 9(3), e92351-e92351 (2014-03-13)
Despite great progress in treatment, the prognosis for patients with esophageal squamous cell carcinoma (ESCC) remains poor, highlighting the importance of early detection. Although upper endoscopy can be used for the screening of esophagus, it has limited sensitivity for early
Chong Da Tan et al.
The Journal of physiology, 592(23), 5251-5268 (2014-09-28)
In cystic fibrosis (CF) lung disease, the absence of functional CF transmembrane conductance regulator results in Cl(-)/HCO3 (-) hyposecretion and triggers Na(+) hyperabsorption through the epithelial Na(+) channel (ENaC), which contribute to reduced airway surface liquid (ASL) pH and volume.
E Campo et al.
The American journal of pathology, 145(2), 301-309 (1994-08-01)
Cathepsin B is a lysosomal cysteine proteinase that has the ability to degrade several extracellular matrix components at both neutral and acidic pH and has been implicated in the progression of several human and rodent tumors. We have studied the
B Werle et al.
Cancer, 89(11), 2282-2291 (2001-01-09)
Tumor cells require specific proteolytic enzymes for invasion and metastasis, including lysosomal peptidases--cathepsins. Cathepsin B is a lysosomal cysteine peptidase, which appears to play a major role in invasion and metastasis of human tumors. In this study, the authors focused
J A Foekens et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 16(3), 1013-1021 (1998-03-21)
Evaluation of the clinical significance of cytosolic tumor levels of the lysosomal cysteine proteases cathepsin B (catB) and cathepsin L (catL) in patients with primary breast cancer. CatB (n = 1,500) and catL (n = 1,391) levels were determined by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service