Skip to Content
Merck
All Photos(3)

Key Documents

HPA017292

Sigma-Aldrich

Anti-B3GNT5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BGnT-5, Anti-Beta-1,3-N- acetylglucosaminyltransferase-5, Anti-Beta3Gn-T5, Anti-Cer synthase, Anti-Lactotriaosylceramide synthase, Anti-Lc(3, Anti-Lc3 synthase, Anti-UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

GAPPIRDKSSKYYVSYEMYQWPAYPDYTAGAAYVISGDVAAKVYEASQTLNSSLYIDDVFMGLCANKIGIVPQDHVFFSGEGKTPYHPCIYEKMMTSHGHLEDLQDLWKNATDPKV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... B3GNT5(84002)

General description

B3GNT5 (UDP-GlcNAc: β Gal β-1,3-N-acetylglucosaminyltransferase 5) is a type II membrane protein belonging to the β3Gn-T family (β3Gn-T5). Its three motif sturcture consists of a NH2-terminal cytoplasmic domain, a transmembrane region, and a stem region and catalytic domain. It is highly expressed in the lung, colon, placenta, testis, pituitary gland. In addition, it is also expressed at low level in cerebellum.

Immunogen

UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

B3GNT5 (UDP-GlcNAc: β Gal β-1,3-N-acetylglucosaminyltransferase 5) plays a key role in the biosynthesis of Lewis antigens. It is mainly responsible for the synthesis of short polylactosamine chains or initiation of polylactosamine synthesis. It has been reported that overexpressed B3GNT5 in human gastric carcinoma cell lines alters sialyl-Lewis x expression and Helicobacter pylori adhesion.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72266

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Togayachi et al.
The Journal of biological chemistry, 276(25), 22032-22040 (2001-04-03)
A new member of the UDP-N-acetylglucosamine:beta-galactose beta1,3-N-acetylglucosaminyltransferase (beta3Gn-T) family having the beta3Gn-T motifs was cloned from rat and human cDNA libraries and named beta3Gn-T5 based on its position in a phylogenetic tree. We concluded that beta3Gn-T5 is the most feasible
Nuno T Marcos et al.
The Journal of clinical investigation, 118(6), 2325-2336 (2008-05-17)
Chronic Helicobacter pylori infection is recognized as a cause of gastric cancer. H. pylori adhesion to gastric cells is mediated by bacterial adhesins such as sialic acid-binding adhesin (SabA), which binds the carbohydrate structure sialyl-Lewis x. Sialyl-Lewis x expression in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service