Skip to Content
Merck
All Photos(6)

Key Documents

HPA017206

Sigma-Aldrich

Anti-CAMK4 antibody produced in rabbit

affinity isolated antibody, Prestige Antibodies® Powered by Atlas Antibodies, buffered aqueous glycerol solution

Synonym(s):

Anti-CAM kinase-GR, Anti-CaMK IV, Anti-Calcium/calmodulin-dependent protein kinase type IV

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

GIITYILLCGFEPFYDERGDQFMFRRILNCEYYFISPWWDEVSLNAKDLVRKLIVLDPKKRLTTFQALQHPWVTGKAANFVHMDTAQKKLQEFNARRKLKAAVKAVVASSRLGSASSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAMK4(814)

Immunogen

Calcium/calmodulin-dependent protein kinase type IV recombinant protein epitope signature tag (PrEST)

Application

Anti-CAMK4 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues.[1] These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting.[1] To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CAMK4 (calcium/calmodulin-dependent protein kinase IV) is a multifunctional serine/threonine protein kinase belonging to the CaMK family. It is a Ca2+ sensor molecule that enters the nucleus via adapter protein, importin-α. It binds to and activates calcium response-element binding (CREB) protein by phosphorylation for gene transcriptional regulation. It can also control alternate splicing by interacting with the ribonuclear protein L. The hippocampus and cerebellum of the CNS (central nervous system) that are responsible for long-term depression (LTD), dendritic growth, and the acquisition of memories are regulated by CAMK4. It also controls the phosphorylation of protamines during spermatogenesis, in terms of, controlling human sperm motility. Thus, a mutation in CAMK4 causes the infertility in men due to abnormal semen activity.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71434

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin J Harrison et al.
The Journal of comparative neurology, 522(2), 308-336 (2013-07-03)
Calcium/calmodulin-dependent protein kinase 4 (gene and transcript: CaMK4; protein: CaMKIV) is the nuclear effector of the Ca(2+) /calmodulin kinase (CaMK) pathway where it coordinates transcriptional responses. However, CaMKIV is present in the cytoplasm and axons of subpopulations of neurons, including
A Khattri et al.
International journal of andrology, 35(6), 810-818 (2012-08-18)
Calcium/calmodulin-dependent protein kinase IV (CAMK4) is a multifunctional serine/threonine protein kinase, which plays an important role in the spermatogenesis by phosphorylating protamines. It has been shown to be involved in the regulation of human sperm motility. Moreover, the Camk4 knockout

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service