Skip to Content
Merck
All Photos(3)

Key Documents

HPA015667

Sigma-Aldrich

Anti-RASGRP2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CalDAG-GEFI, Anti-Calcium and DAG-regulated guanine nucleotide exchange factor I, Anti-Cdc25-like protein, Anti-F25B33 kinase-like protein, Anti-HCDC25L, Anti-RAS guanyl-releasing protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Estimated to ship on29 April 2025



Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Estimated to ship on29 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

DDSGKVRDPQLVRMFLMMHPWYIPSSQLAAKLLHIYQQSRKDNSNSLQVKTCHLVRYWISAFPAEFDLNPELAEQIKELKALLDQEGNRRHSSLIDIDSVPTYKWKRQVTQRNPV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RASGRP2(10235)

Immunogen

RAS guanyl-releasing protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RAS guanyl-releasing protein 2 (a calcium- and diacylglycerol-responsive guanine nucleotide exchange factor) is a protein encoded by the RASGRP2 gene in humans. It encodes for calcium- and DAG-regulated guanine exchange factor-1 (CalDAG-GEFI), which acts as a novel candidate for preventing thrombosis. CalDAG-GEFI is found to be expressed mainly in the striatum and olfactory structures and deep cortical layers and may serve as a key intracellular regulators. In endothelial cells, it increases the viability of cells and cell-matrix adhesion by increasing Ras expression and Rap1 activation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72501

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junichi Takino et al.
Cell adhesion & migration, 7(3), 262-266 (2013-04-09)
Ras guanyl nucleotide releasing proteins (RasGRPs) are guanine nucleotide exchange factors that activate Ras and Rap. We recently reported that xrasgrp2, which is a homolog of the human rasgrp2, plays a role in vasculogenesis and/or angiogenesis during early development of
Kentaro Nagamine et al.
Biological & pharmaceutical bulletin, 33(7), 1138-1142 (2010-07-08)
Ras guanyl nucleotide-releasing protein 2 (RASGRP2) is a calcium- and diacylglycerol-responsive guanine nucleotide exchange factor. Previously, we reported that XRASGRP2, a homolog of human RASGRP2, is expressed in the vascular region of the Xenopus embryo during embryogenesis. Here, we report
S Toki et al.
The Journal of comparative neurology, 437(4), 398-407 (2001-08-15)
CalDAG-GEFI and CalDAG-GEFII (identical to RasGRP) are novel, brain-enriched guanine nucleotide exchange factors (GEFs) that can be stimulated by calcium and diacylglycerol and that can activate small GTPases, including Ras and Rap1, molecules increasingly recognized as having signaling functions in
Human CalDAG-GEFI gene (RASGRP2) mutation affects platelet function and causes severe bleeding.
Canault M, Ghalloussi D, Grosdidier C, et al.
The Journal of Experimental Medicine, 211(7), 1349-1362 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service