Skip to Content
Merck
All Photos(3)

Key Documents

HPA015024

Sigma-Aldrich

Anti-GJC3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Connexin-30.2, Anti-Connexin-31.3, Anti-Cx30.2, Anti-Cx31.3, Anti-Gap junction γ-3 protein, Anti-Gap junction ε-1 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20-1:50
western blot: 0.04-0.4 μg/mL

immunogen sequence

RTWKHKSSSSKYFLTSESTRRHKKATDSLPVVETKEQFQEAVPGRSLAQEKQRPVGPRDA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GJC3(349149)

General description

GJC3 (gap junction protein, γ 3) belongs to the family of four-transmembrane domain containing gap junction channels called connexins. It is also called CX30.2; CX31.3. It is predominantly expressed in heart, liver and skeletal muscle, and is rarely expressed in brain, kidney and spinal cord. This gene is localized to human chromosome 7q22.1, contains two exons, and codes for a protein of 31.29kDa, which is composed of 279 amino acids.

Immunogen

Gap junction γ-3 protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GJC3 (gap junction protein, γ 3) forms a component of gap junctions (GJs), and these GJs help in intracellular communication, and maintain tissue homeostasis. Mutations in this gene lead to the loss of its hemichannel function, which results in non-syndromic hearing loss. Studies show that this protein is essential for normal neuronal and hearing development. In HeLa cells stably expressing GJC3, there is elevated release of ATP.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73074

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ching-Chyuan Su et al.
Cell biochemistry and biophysics, 66(2), 277-286 (2012-11-28)
Connexins (CXs), as a component of gap junction channel, are homologous four transmembrane-domain proteins, with numerous studies confirming their auditory functions. Among a cohort of patients having incurred non-syndromic hearing loss, we identified two novel missense mutations, p.R15G and p.L23H
Wei-Guang Liang et al.
Cell biochemistry and biophysics, 61(1), 189-197 (2011-04-12)
Gap junctional intercellular communication has numerous functions, each of which meets the particular needs of organs, tissues, or groups of cells. Connexins (CXs) are homologous four-transmembrane-domain proteins that are the major components of gap junctions. CX30.2/CX31.3 (GJC3) is a relatively

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service