Skip to Content
Merck
All Photos(4)

Key Documents

HPA014963

Sigma-Aldrich

Anti-SLC13A2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Na(+/dicarboxylate cotransporter 1, Anti-Renal sodium/dicarboxylate cotransporter, Anti-Solute carrier family 13 member 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

ATSAMMVPIAHAVLDQLHSSQASSNVEEGSNNPTFELQEPSPQKEVTKLDNGQALPVTSASSEGRAHLSQKHLHLTQCMSLCVCYSASIGGIATLTGTAPNLVLQGQINSLFPQNGNVV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC13A2(9058)

General description

SLC13A2 (solute carrier family 13, member 2) is a member of the SLC13 family which contains five members. It is a Na+-dicarboxylate cotransporter, present in the kidney. It has two N-glycosylation sites present on its C-terminal. Apart from kidney, the mRNA is also found in intestine. It is present on proximal tubular cells, on their apical sides. It is also called Na+-coupled dicarboxylate transporters 1 (NaDC1), and is one of the two isoforms of NaDC transporters. This gene is present on human chromosome 17, spans ~30kb, and has around 12 coding exons.

Immunogen

Solute carrier family 13 member 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SLC13A2 (solute carrier family 13, member 2) is essential for proper citrate excretion, and reabsorbs divalent citrate (citrate2-) at physiological pH, present as ~10% of the free plasma citrate. Urinary citrate complexes Ca2+ ions, and thus prevents nephrolithiasis. Thus, this gene might be associated with the pathophysiology of nephrolithiasis. It is over-expressed in metabolic acidosis, leading to hypocitraturia. Its expression is also down-regulated in metabolic alkalosis. It acts as a transporter for succinate and other dicarboxylates, but with low-affinity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73200

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc J Bergeron et al.
The Journal of biological chemistry, 286(13), 11242-11253 (2011-01-25)
Renal excretion of citrate, an inhibitor of calcium stone formation, is controlled mainly by reabsorption via the apical Na(+)-dicarboxylate cotransporter NaDC1 (SLC13A2) in the proximal tubule. Recently, it has been shown that the protein phosphatase calcineurin inhibitors cyclosporin A (CsA)
Ana M Pajor
Pflugers Archiv : European journal of physiology, 451(5), 597-605 (2005-10-08)
The SLC13 gene family consists of five members in humans, with corresponding orthologs from different vertebrate species. All five genes code for sodium-coupled transporters that are found on the plasma membrane. Two of the transporters, NaS1 and NaS2, carry substrates
A M Pajor
The American journal of physiology, 270(4 Pt 2), F642-F648 (1996-04-01)
The renal Na(+)-dicarboxylate cotransporter reabsorbs Krebs cycle intermediates, such as succinate and citrate, from the glomerular filtrate. The present study describes the cloning and characterization of the human renal Na(+)-dicarboxylate cotransporter, hNaDC-1. The amino acid sequence of hNaDC-1 is 78%
Katsuhisa Inoue et al.
Biochemical and biophysical research communications, 299(3), 465-471 (2002-11-26)
This paper describes the cloning and functional characterization of the human Na(+)-coupled citrate transporter (NaCT). The cloned human NaCT shows 77% sequence identity with rat NaCT. The nact gene is located on human chromosome 17 at p12-13. NaCT mRNA is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service