Skip to Content
Merck
All Photos(4)

Key Documents

HPA014703

Sigma-Aldrich

Anti-CLDN7 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CLDN-7, Anti-Claudin-7

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

PQWQMSSYAGDNIITAQAMYKGLWMDCVTQSTGMMSCKMYDSVLALS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLDN7(1366)

General description

CLDN7 (claudin-7) belongs to the family of tight junction proteins called claudins, which are responsible for the maintenance of epithelial barrier function. This family has at least 24 members, and they are all membrane proteins. It has a short N-terminal facing the cytosol, two extraplasmic domains, and cytosolic C-terminal. Its spans the membrane four times.

Immunogen

Claudin-7 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CLDN7 (claudin-7) prevents the invasion and migration of human lung cancer cells, and induces apoptosis by activating caspases, post cisplatin treatment of lung cancer cells. It interacts with CD44 and EpCAM, and might form a membrane-anchoring protein complex, which helps it to reside in the plasma membrane. It is up-regulated in colon cancer, where it facilitates tumorigenesis. Its expression is also associated with inflammatory bowel disease (IBD) and colitis-associated cancer. It is either completely inactivated or under-expressed in human breast cancer, and is also linked with relapse of the same. Studies on human breast cancer MCF-7 cell line, show that tumor necrosis factor (TNF)α induces the expression of CLDN7, which leads to accumulation of β-catenin in the cytosol. This way CLDN7 regulates apoptosis of MCF-7 cell line. It is over-expressed in epithelial ovarian cancer, where it might be involved in its invasion.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73174

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John Hoggard et al.
Cancer science, 104(5), 611-618 (2013-02-26)
Claudins are a family of tight junction (TJ) integral membrane proteins that play a crucial role in maintaining cell polarity, adhesion, and paracellular permeability. Changes in expression levels of claudin proteins have been associated with human lung cancer. Previously, we
Jillian L Pope et al.
Molecular cancer, 13, 167-167 (2014-07-07)
The tight junction protein Claudin-1, a claudin family member, has been implicated in several gastro-intestinal pathologies including inflammatory bowel disease (IBD) and colorectal cancer (CRC). In this regard, we have demonstrated that claudin-1 expression in colon cancer cells potentiates their
Neetu Dahiya et al.
PloS one, 6(7), e22119-e22119 (2011-07-27)
Claudins are tight junction proteins that are involved in tight junction formation and function. Previous studies have shown that claudin-7 is frequently upregulated in epithelial ovarian cancer (EOC) along with claudin-3 and claudin-4. Here, we investigate in detail the expression
Yang Liu et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 33(6), 2307-2315 (2012-09-04)
Accumulating evidence reveals that aberrant expression of claudins manifests in various tumors; however, their biological functions are poorly understood. Here, we report on the elevated expression of claudin-1 in human breast cancer MCF-7 cells under tumor necrosis factor (TNF)-α treatment.
Hiroki Fujita et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 54(8), 933-944 (2006-05-03)
Among tight-junction proteins, claudins, which play a key role in paracellular transport across epithelia, claudins 1 to 5 are expressed in the intestine, and changes in their abundance and/or distribution are considered to contribute to various gastrointestinal diseases. We investigated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service