Skip to Content
Merck
All Photos(7)

Key Documents

HPA014492

Sigma-Aldrich

Anti-PDE3A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CGI-PDE A, Anti-Cyclic GMP-inhibited phosphodiesterase A, Anti-cGMP-inhibited 3′,5′-cyclic phosphodiesterase A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SPQILTPPVICSSCGRPYSQGNPADEPLERSGVATRTPSRTDDTAQVTSDYETNNNSDSSDIVQNEDETECLREPLRKASACSTYAPETMMFLDKPILAPEPLVMDNLDSIMEQLNT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDE3A(5139)

General description

The gene PDE3A (phosphodiesterase 3A) encodes a cyclic nucleotide phosphodiesterase that is expressed mainly in cardiac and vascular myocytes and platelets. The cDNA contains a C-terminal catalytic region that spans around 280 amino acids and two hydrophobic regions at the N-terminus that participate in intracellular targeting. The gene containing 16 exons spanning over 130kb is mapped to human chromosome 12p12.

Immunogen

cGMP-inhibited 3′,5′-cyclic phosphodiesterase A recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The cyclic nucleotide PDEs (phosphodiesterases) hydrolyze cyclic nucleotides, cAMP and cGMP. The isoforms of PDE3 are cGMP-inhibited phosphodiesterases that hydrolyze cAMP and participate in the regulation of myocardial contractility, aggregation of platelets, and vascular smooth muscle relaxation. PDE3A activity is crucial for the maintenance of low equilibrium levels of cAMP and to determine the onset for platelet activation. Its activity can be increased by PKC (protein kinase C)-mediated phosphorylation, in turn regulating cAMP levels in human platelets.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72629

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roger W Hunter et al.
The Journal of biological chemistry, 284(18), 12339-12348 (2009-03-06)
The elevation of [cAMP](i) is an important mechanism of platelet inhibition and is regulated by the opposing activity of adenylyl cyclase and phosphodiesterase (PDE). In this study, we demonstrate that a variety of platelet agonists, including thrombin, significantly enhance the
Kirsi Toivanen et al.
Cancers, 15(22) (2023-11-25)
Liposarcomas (LPSs) are a heterogeneous group of malignancies that arise from adipose tissue. Although LPSs are among the most common soft-tissue sarcoma subtypes, precision medicine treatments are not currently available. To discover LPS-subtype-specific therapy targets, we investigated RNA sequenced transcriptomes
Jeremy Wechsler et al.
The Journal of biological chemistry, 277(41), 38072-38078 (2002-08-03)
PDE3A cyclic nucleotide phosphodiesterases regulate cAMP- and cGMP-mediated intracellular signaling in cardiac myocytes. We used antibodies to different regions of PDE3A to demonstrate the presence of three PDE3A isoforms in these cells. These isoforms, whose apparent molecular weights are 136,000
J Kasuya et al.
Biochemical and biophysical research communications, 268(3), 827-834 (2000-02-19)
Phosphodiesterase type 3 isoforms, PDE3A and 3B, are expressed primarily in cardiovascular and adipose tissues, respectively. We previously reported a shorter transcript of 4.4-kb PDE3A which is predominantly transcribed in human placenta, whereas a full-length 7. 6-kb transcript corresponding to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service