Skip to Content
Merck
All Photos(2)

Key Documents

HPA014377

Sigma-Aldrich

Anti-SIGLEC14 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

CQVKRQGAQVTTERTVQLNVSYAPQNLAISIFFRNGTGTALRILSNGMSVPIQEGQSLFLACTVDSNPPASLSWFREGKALNPSQTSMSGTLELPNIGAREGGEFTCRVQHPLGSQHLSF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The gene SIGLEC14 (sialic acid binding Ig-like lectin 14) encodes a member of the Siglec family of sialic acid-binding lectins. It is found to be expressed mainly on granulocytes and monocytes. Human Siglec-14 is very similar to human Siglec-5 in terms of the sequence. The members of this family are receptors of the immunoglobulin (Ig) superfamily and are expressed on immune cells. The encoded protein has three Ig-like domains and preferentially binds to glycan. It also interacts with the activating adapter protein DAP12. The N-terminal region in both the proteins is almost identical and this indicates partial gene conversion between SIGLEC14 and SIGLEC5. The essential amino acid arginine of this protein, that participates in the recognition of sialic acid, is present in humans and is mutated in other apes.

Immunogen

Sialic acid-binding Ig-like lectin 14 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Siglec-14 (sialic acid binding Ig-like lectin 14) enhances the secretion of TNF-α (tumor necrosis fector) stimulated by lipopolysaccharide in a monocytic cell line, indicating its role in bacterial infection. It is also involved in the ′infection-inflammation-exacerbation′ axis of COPD (chronic obstructive pulmonary disease) progression.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72436

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Angata et al.
Cellular and molecular life sciences : CMLS, 70(17), 3199-3210 (2013-03-23)
Chronic obstructive pulmonary disease (COPD) is a leading cause of mortality worldwide. COPD exacerbation, or episodic worsening of symptoms, often results in hospitalization and increased mortality rates. Airway infections by new bacterial strains, such as nontypeable Haemophilus influenzae (NTHi), are
Masahiro Yamanaka et al.
Glycobiology, 19(8), 841-846 (2009-04-17)
Human Siglec-14, a member of the Siglec family of sialic acid-binding lectins, shows extensive sequence similarity to human Siglec-5. To analyze respective expression patterns of Siglec-14 and Siglec-5, we developed specific antibodies against each of them. We found that the
Takashi Angata et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 20(12), 1964-1973 (2006-10-03)
Immune receptors that show high mutual sequence similarity and have antagonizing signaling properties are called paired receptors, and are believed to fine-tune immune responses. Siglecs are sialic acid-recognizing receptors of the immunoglobulin (Ig) superfamily expressed on immune cells. Human Siglec-5

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service