Skip to Content
Merck
All Photos(6)

Key Documents

HPA014044

Sigma-Aldrich

Anti-CLEC4G antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C-type lectin domain family 4 member G

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

QGFLTRNTRGRGYWLGLRAVRHLGKVQGYQWVDGVSLSFSHWNQGEPNDAWGRENCVMMLHTGLWNDAPCDSEKDGWICEKR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLEC4G(339390)

General description

The gene CLEC4G (C-type lectin domain family 4 member G) gene forms a lectin gene cluster at human chromosome 19p13.3 along with DC-SIGN, and L-SIGN and CD23. CLEC4G is expressed on sinusoidal endothelial cells in liver and lymph nodes along with DC-SIGNR. The C-type lectins contain a C-terminal carbohydrate recognition domain (CRD) that functions as an attachment factor. This domain links high mannose oligosaccharides or N-linked glycosylated protein. The neck region of CRD contains repeat units called the VNTR (variable number tandem repeat).

Immunogen

C-type lectin domain family 4 member G recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The C-type lectin encoded by the gene CLEC4G (C-type lectin domain family 4 member G) plays an important role in the activation of B-cells. It interacts with and mediates viral and bacterial pathogen recognitions. The human CLEC4G protein was previously referred to as LSECtin. It is capable of interacting with BACE1 (β-Site amyloid precursor protein cleaving enzyme-1 ) molecule and negatively regulate it. BACE1 cleaves amyloid precursor protein (APP) to form toxic amyloid-β (Aβ) peptides and has been implicated in Alzheimer′s disease (AD).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72322

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y W Huang et al.
Developmental and comparative immunology, 33(6), 747-760 (2009-01-27)
Human CLEC4G (previously named LSECtin), DC-SIGN, and L-SIGN are three important C-type lectins capable of mediating viral and bacterial pathogen recognitions. These three genes, together with CD23, form a lectin gene cluster at chromosome 19p13.3. In this study, we have
H Li et al.
Journal of medical genetics, 45(11), 752-758 (2008-08-14)
Polymorphisms of CLEC4M have been associated with predisposition for infection by the severe acute respiratory syndrome coronavirus (SARS-CoV). DC-SIGNR, a C-type lectin encoded by CLEC4M, is a receptor for the virus. A variable number tandem repeat (VNTR) polymorphism in its
Yasuhiko Kizuka et al.
FEBS letters, 589(13), 1418-1422 (2015-05-11)
β-Site amyloid precursor protein cleaving enzyme-1 (BACE1) is a central molecule in Alzheimer's disease (AD). It cleaves amyloid precursor protein (APP) to produce the toxic amyloid-β (Aβ) peptides. Thus, a novel BACE1 modulator could offer a new therapeutic strategy for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service