Skip to Content
Merck
All Photos(10)

Key Documents

HPA012720

Sigma-Aldrich

Anti-PPIB antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CYP-S1, Anti-Cyclophilin B, Anti-PPIase, Anti-Peptidyl-prolyl cis-trans isomerase B precursor, Anti-Rotamase, Anti-S-cyclophilin, Anti-SCYLP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Check Cart for Availability


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

enhanced validation

RNAi knockdown
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

LKHYGPGWVSMANAGKDTNGSQFFITTVKTAWLDGKHVVFGKVLEGMEVVRKVESTKTDSRDKPLKDVIIADCGKIEVEKPFAIAKE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPIB(5479)

General description

Peptidyl -prolyl cis-trans isomerase B (PPIB) is a cyclosporin A binding protein localized to the calcium-containing intracellular vesicles. It is expressed in the endoplasmic reticulum and nuclear matrix. The gene encoding this protein is present on chromosome 15.

Immunogen

Peptidyl-prolyl cis-trans isomerase B precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-PPIB antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting.† To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Western Blotting (1 paper)

Biochem/physiol Actions

Peptidyl -prolyl cis-trans isomerase B (PPIB) is involved in the activation of peripheral blood T-lymphocytes and has the ability to mediate immunosuppressant Cyclosporin A inhibition. It interacts with growth hormone and prolactin, and acts as a chaperone facilitating their nuclear retro-transport.[1] Mutations in the gene cause severe osteogenesis imperfecta.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72173

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of cyclophilins in somatolactogenic action.
MA Rycyzyn and CV Clevenger
Annals of the New York Academy of Sciences, 917, 514-521 (2000)
Identification of the immunophilins capable of mediating inhibition of signal transduction by cyclosporin A and FK506: roles of calcineurin binding and cellular location.
Bram RJ, et al.
Molecular and Cellular Biology, 13(8), 4760-4769 (1993)
Takuo Hayashi et al.
PloS one, 8(12), e81883-e81883 (2013-12-18)
Metastasis is still a major issue in cancer, and the discovery of biomarkers predicting metastatic capacity is essential for the development of better therapeutic strategies for treating lung adenocarcinoma. By using a proteomic approach, we aimed to identify novel predictors
L B Peddada et al.
Cytogenetics and cell genetics, 60(3-4), 219-221 (1992-01-01)
The polymerase chain reaction (PCR) technique was used to generate a unique probe complementary to the hydrophobic 5' end of the human cyclophilin B gene. This unique probe was hybridized to DNAs from human x hamster hybrid somatic cell lines
M Le Hir et al.
Laboratory investigation; a journal of technical methods and pathology, 73(5), 727-733 (1995-11-01)
The immunosuppressant cyclosporin A (CsA) forms a trimolecular complex with cyclophilin (CPH) and calcineurins (CN) and inhibits CN phosphatase activity. Inhibition of CN phosphatase by CsA prevents the dephosphorylation of a nuclear factor in the cytosol and its nuclear translocation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service